Gene Transcripts

Gene Symbol
SCGB1D2
Entrez Gene ID
10647
Gene Name
secretoglobin family 1D member 2
Gene Aliases
LIPB, LPHB, LPNB
Chromosome Location
Chr. 11: 62242252 - 62244808 on Build GRCh38
UniGene ID
Hs.204096
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006551.3 NP_006542.1 2 154
GenBank mRNA AJ224172.1 - 2 180
AL708956.1 - 2 179
BC069290.1 - 2 148 7262705
BC104838.1 - 2 124 8143841
BC104840.1 - 2 123 8143843

View Full Product Details