Gene Transcripts

Gene Symbol
HMGN5
Entrez Gene ID
79366
Gene Name
high mobility group nucleosome binding domain 5
Gene Aliases
NBP-45, NSBP1
Chromosome Location
Chr. X: 81113701 - 81201942 on Build GRCh38
UniGene ID
Hs.282204
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030763.2 NP_110390.1 6 549
XM_006724688.3 XP_006724751.1 6 499
XM_017029839.1 XP_016885328.1 4 379
XM_017029840.1 XP_016885329.1 4 373
GenBank mRNA AF250329.1 - 5 288
AK094058.1 - 6 549
AK315792.1 - 5 342
BC005342.1 - 5 309 3838879
DA745749.1 - 6 518

View Full Product Details