Gene Transcripts

Gene Symbol
UBL4B
Entrez Gene ID
164153
Gene Name
ubiquitin like 4B
Gene Aliases
Chromosome Location
Chr. 1: 110112440 - 110113947 on Build GRCh38
UniGene ID
Hs.374027
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203412.1 NP_981957.1 1 320
GenBank mRNA AK098556.1 - 1 321
BC058929.1 - 1 292 5741847

View Full Product Details