Gene Transcripts

Gene Symbol
MAMSTR
Entrez Gene ID
284358
Gene Name
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases
MASTR
Chromosome Location
Chr. 19: 48705718 - 48719898 on Build GRCh38
UniGene ID
Hs.191815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130915.1 NP_001124387.1 9 1011
NM_001297753.1 NP_001284682.1 6 1124
NM_182574.2 NP_872380.1 7 1255
XM_011526807.2 XP_011525109.1 7 1260
XM_011526808.2 XP_011525110.1 9 1379
XM_011526809.2 XP_011525111.1 6 1058
XM_017026640.1 XP_016882129.1 8 1779
XM_017026641.1 XP_016882130.1 8 1740
XM_017026643.1 XP_016882132.1 8 1620
XM_017026644.1 XP_016882133.1 8 1590
XM_017026645.1 XP_016882134.1 5 1025
GenBank mRNA AK093389.1 - 7 1255
AK131427.1 - 9 1018
BC105056.1 - 5 679 8144059
BC105058.1 - 5 679 8144061
BC106047.1 - 6 1124 5786639
BC143443.1 - 4 484 9051951
BX280379.1 - 2 205

View Full Product Details