Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145028.1 | NP_001138500.1 | 4 | 295 | |
XM_005259891.4 | XP_005259948.1 | 5 | 496 | ||
XM_011527965.2 | XP_011526267.1 | 4 | 416 | ||
GenBank mRNA | AW130597.1 | - | 2 | 273 | |
CR747190.1 | - | 3 | 418 |