Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001134649.2 | NP_001128121.1 | 7 | 668 | |
NM_001134650.1 | NP_001128122.1 | 7 | 544 | ||
NM_001134651.1 | NP_001128123.1 | 6 | 535 | ||
NM_001282886.1 | NP_001269815.1 | 7 | 661 | ||
NM_173359.4 | NP_775495.1 | 7 | 629 | ||
XM_011533651.2 | XP_011531953.1 | 4 | 388 | ||
XM_017006278.1 | XP_016861767.1 | 8 | 675 | ||
XM_017006279.1 | XP_016861768.1 | 8 | 662 | ||
XM_017006280.1 | XP_016861769.1 | 8 | 809 | ||
XM_017006281.1 | XP_016861770.1 | 6 | 537 | ||
XM_017006282.1 | XP_016861771.1 | 8 | 617 | ||
GenBank mRNA | AK126999.1 | - | 6 | 417 | |
AK313656.1 | - | 7 | 629 | ||
BC031289.2 | - | 5 | 352 | 5299499 | |
BC068443.1 | - | 7 | 661 | 4820931 | |
BX648727.1 | - | 3 | 149 | ||
DA227887.1 | - | 7 | 519 | ||
DA394405.1 | - | 7 | 544 | ||
DR159502.1 | - | 6 | 534 |