Gene Transcripts
- Gene Symbol
- GSTM2P1
- Entrez Gene ID
- 442245
- Gene Name
- glutathione S-transferase mu 2 pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 111046419 - 111047554 on Build GRCh38
- UniGene ID
- Hs.551627
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002932.2 |
|
1 |
348 |
|
GenBank mRNA |
AY532924.1 |
- |
1 |
315 |
|
AY532925.1 |
- |
1 |
315 |
|
View Full Product Details