Gene Transcripts
- Gene Symbol
- CALML3-AS1
- Entrez Gene ID
- 100132159
- Gene Name
- CALML3 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 10: 5514244 - 5526246 on Build GRCh38
- UniGene ID
- Hs.239600
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_120496.1 |
|
4 |
2009 |
|
NR_120497.1 |
|
3 |
1897 |
|
GenBank mRNA |
AK128534.1 |
- |
3 |
2423 |
|
View Full Product Details