Gene Transcripts

Gene Symbol
SNRNP27
Entrez Gene ID
11017
Gene Name
small nuclear ribonucleoprotein U4/U6.U5 subunit 27
Gene Aliases
27K, RY1
Chromosome Location
Chr. 2: 69893943 - 69905236 on Build GRCh38
UniGene ID
Hs.54649
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006857.2 NP_006848.1 2 155
NR_037862.1 2 155
GenBank mRNA AK291128.1 - 2 136
AK312041.1 - 2 120
BC017890.1 - 2 140 4699198
BU171817.1 - 2 146
X76302.1 - 2 139

View Full Product Details