Gene Transcripts

Gene Symbol
CTRC
Entrez Gene ID
11330
Gene Name
chymotrypsin C
Gene Aliases
CLCR, ELA4
Chromosome Location
Chr. 1: 15438442 - 15449247 on Build GRCh38
UniGene ID
Hs.631869
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007272.2 NP_009203.2 6 648
GenBank mRNA AK289447.1 - 6 647
BC015118.1 - 6 654 3950170
S82198.1 - 6 625

View Full Product Details