Gene Transcripts
- Gene Symbol
- FCAR
- Entrez Gene ID
- 2204
- Gene Name
- Fc fragment of IgA receptor
- Gene Aliases
- CD89, CTB-61M7.2, FcalphaRI
- Chromosome Location
- Chr. 19: 54874093 - 54891420 on Build GRCh38
- UniGene ID
- Hs.659872
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002000.3 |
NP_001991.1
|
4, 5 |
851 |
|
NM_133272.3 |
NP_579806.1
|
3, 4 |
815 |
|
NM_133277.3 |
NP_579811.1
|
2, 3 |
524 |
|
XM_011526625.2 |
XP_011524927.1
|
4, 5 |
775 |
|
XM_017026473.1 |
XP_016881962.1
|
4, 5 |
824 |
|
XM_017026474.1 |
XP_016881963.1
|
3, 4 |
607 |
|
GenBank mRNA |
AK290922.1 |
- |
4, 5 |
709 |
|
BC027953.1 |
- |
4, 5 |
678 |
5211453 |
D87855.1 |
- |
3, 4 |
618 |
|
D87861.1 |
- |
2, 3 |
327 |
|
DQ075334.1 |
- |
4, 5 |
654 |
|
DQ075335.1 |
- |
4, 5 |
654 |
|
DQ075336.1 |
- |
4, 5 |
654 |
|
DQ075337.1 |
- |
4, 5 |
654 |
|
DQ075338.1 |
- |
4, 5 |
654 |
|
DQ075339.1 |
- |
4, 5 |
654 |
|
X92558.1 |
- |
3, 4 |
618 |
|
X92562.1 |
- |
2, 3 |
327 |
|
View Full Product Details