Gene Transcripts
- Gene Symbol
- TCAIM
- Entrez Gene ID
- 285343
- Gene Name
- T-cell activation inhibitor, mitochondrial
- Gene Aliases
- C3orf23, TOAG-1, TOAG1
- Chromosome Location
- Chr. 3: 44338119 - 44409448 on Build GRCh38
- UniGene ID
- Hs.55131
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001029839.2 |
NP_001025010.1
|
2, 3 |
447 |
|
NM_001029840.2 |
NP_001025011.1
|
2, 3 |
265 |
|
NM_001282913.1 |
NP_001269842.1
|
2, 3 |
265 |
|
NM_001282914.1 |
NP_001269843.1
|
2, 3 |
447 |
|
NM_001282915.1 |
NP_001269844.1
|
2, 3 |
265 |
|
NM_173826.3 |
NP_776187.2
|
2, 3 |
447 |
|
XM_017006251.1 |
XP_016861740.1
|
2, 3 |
492 |
|
GenBank mRNA |
AK056593.1 |
- |
2, 3 |
447 |
|
AK292756.1 |
- |
2, 3 |
265 |
|
AL832473.1 |
- |
2, 3 |
124 |
|
BC009400.2 |
- |
2, 3 |
243 |
|
BC038420.1 |
- |
2, 3 |
367 |
|
BC047231.1 |
- |
2, 3 |
142 |
4817904 |
BC062587.1 |
- |
2, 3 |
419 |
|
BC073829.1 |
- |
2, 3 |
92 |
|
BC101058.2 |
- |
2, 3 |
124 |
|
BC101059.2 |
- |
2, 3 |
124 |
40008143 |
BC101060.2 |
- |
2, 3 |
124 |
40008144 |
BC101061.2 |
- |
2, 3 |
124 |
40008152 |
CN372173.1 |
- |
2, 3 |
220 |
|
DC413255.1 |
- |
2, 3 |
265 |
|
View Full Product Details