Gene Transcripts

Gene Symbol
ACACA
Entrez Gene ID
31
Gene Name
acetyl-CoA carboxylase alpha
Gene Aliases
ACAC, ACACAD, ACC, ACC1, ACCA
Chromosome Location
Chr. 17: 37084992 - 37406822 on Build GRCh38
UniGene ID
Hs.160556
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198834.2 NP_942131.1 21, 22 3244
NM_198836.2 NP_942133.1 21, 22 2812
NM_198837.1 NP_942134.1 19, 20 2882
NM_198838.1 NP_942135.1 20, 21 2993
NM_198839.2 NP_942136.1 25, 26 3918
XM_005257267.4 XP_005257324.1 21, 22 2725
XM_006721853.1 XP_006721916.1 21, 22 3255
XM_011524701.1 XP_011523003.1 21, 22 3165
XM_011524703.1 XP_011523005.1 21, 22 2797
XM_011524704.2 XP_011523006.1 21, 22 2730
XM_017024553.1 XP_016880042.1 22, 23 3279
XM_017024554.1 XP_016880043.1 22, 23 3279
XM_017024555.1 XP_016880044.1 21, 22 2878
GenBank mRNA AB371587.1 - 21, 22 2791
AK309084.1 - 21, 22 2800
AY237919.1 - 19, 20 2621
AY315627.1 - 19, 20 2621
BC137287.1 - 21, 22 2749 9021284
U19822.1 - 19, 20 2621

View Full Product Details