Gene Transcripts

Gene Symbol
LIMK1
Entrez Gene ID
3984
Gene Name
LIM domain kinase 1
Gene Aliases
LIMK, LIMK-1
Chromosome Location
Chr. 7: 74083777 - 74122525 on Build GRCh38
UniGene ID
Hs.647035
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002314.3 NP_002305.1 2, 3 354
GenBank mRNA AF134379.1 - 2, 3 140
AK290162.1 - 3, 4 279
D26309.1 - 3, 4 235

View Full Product Details