Gene Transcripts
- Gene Symbol
- COMMD10
- Entrez Gene ID
- 51397
- Gene Name
- COMM domain containing 10
- Gene Aliases
- PTD002
- Chromosome Location
- Chr. 5: 116084592 - 116446462 on Build GRCh38
- UniGene ID
- Hs.483136
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001308080.1 |
NP_001295009.1
|
2, 3 |
238 |
|
NM_016144.3 |
NP_057228.1
|
2, 3 |
211 |
|
XM_011543458.2 |
XP_011541760.1
|
2, 3 |
595 |
|
XM_011543460.2 |
XP_011541762.1
|
2, 3 |
594 |
|
XM_017009553.1 |
XP_016865042.1
|
2, 3 |
333 |
|
XM_017009555.1 |
XP_016865044.1
|
2, 3 |
593 |
|
XM_017009556.1 |
XP_016865045.1
|
2, 3 |
593 |
|
GenBank mRNA |
AF078857.1 |
- |
2, 3 |
166 |
|
AF161423.1 |
- |
2, 3 |
162 |
|
AK002147.1 |
- |
2, 3 |
187 |
|
AY542165.1 |
- |
2, 3 |
134 |
|
BC005179.1 |
- |
2, 3 |
157 |
3683093 |
BI859871.1 |
- |
2, 3 |
227 |
|
DA001572.1 |
- |
2, 3 |
238 |
|
HY139221.1 |
- |
2, 3 |
212 |
|
View Full Product Details