Gene Transcripts

Gene Symbol
PROK1
Entrez Gene ID
84432
Gene Name
prokineticin 1
Gene Aliases
EGVEGF, PK1, PRK1
Chromosome Location
Chr. 1: 110451166 - 110457354 on Build GRCh38
UniGene ID
Hs.514793
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032414.2 NP_115790.1 3 350
GenBank mRNA AF333024.1 - 3 353
AY029225.1 - 3 347
AY358683.1 - 3 389
BC025399.1 - 3 327 4826496
BM544315.1 - 3 376

View Full Product Details