Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74457207-74457703 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNE3
Entrez Gene ID
10008
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 3
Gene Aliases
HOKPP, HYPP, MiRP2
UniGene
Hs.523899
Gene Chromosome Location
Chr.11: 74454841-74467729 on Build GRCh38
Transcripts
NM_005472.4 XM_011544713.2 XM_017017047.1 XM_017017048.1 XM_017017049.1 XM_017017050.1 XM_017017051.1 XM_017017052.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6690770 c.15T>C p.N5N Chr.11: 74457549-74457549 on Build GRCh38
6318635 c.295C>A p.R99S Chr.11: 74457269-74457269 on Build GRCh38
6838041 c.97C>T p.P33S Chr.11: 74457467-74457467 on Build GRCh38
1318090 c.83A>G p.N28S Chr.11: 74457481-74457481 on Build GRCh38
6690771 c.291G>T p.K97N Chr.11: 74457273-74457273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775009123 Chr.11: 74457582-74457582 on Build GRCh38
rs1057518549 Chr.11: 74457575-74457575 on Build GRCh38
rs745645715 Chr.11: 74457469-74457469 on Build GRCh38
rs780882536 Chr.11: 74457460-74457460 on Build GRCh38
rs377606406 Chr.11: 74457574-74457574 on Build GRCh38
rs780971201 Chr.11: 74457397-74457397 on Build GRCh38
rs746750241 Chr.11: 74457300-74457300 on Build GRCh38
rs773287275 Chr.11: 74457515-74457515 on Build GRCh38
rs786205809 Chr.11: 74457284-74457284 on Build GRCh38
rs772286478 Chr.11: 74457481-74457481 on Build GRCh38
rs758642757 Chr.11: 74457569-74457569 on Build GRCh38
rs752765414 Chr.11: 74457524-74457524 on Build GRCh38
rs369134829 Chr.11: 74457391-74457391 on Build GRCh38
rs776896849 Chr.11: 74457521-74457521 on Build GRCh38
rs918843154 Chr.11: 74457512-74457512 on Build GRCh38
rs759641427 Chr.11: 74457497-74457497 on Build GRCh38
rs763157740 Chr.11: 74457302-74457302 on Build GRCh38
rs757890580 Chr.11: 74457442-74457442 on Build GRCh38
rs777182014 Chr.11: 74457581-74457581 on Build GRCh38
rs779362496 Chr.11: 74457526-74457526 on Build GRCh38
rs771425703 Chr.11: 74457520-74457520 on Build GRCh38
rs991112634 Chr.11: 74457654-74457654 on Build GRCh38
rs958007485 Chr.11: 74457649-74457649 on Build GRCh38
rs770147077 Chr.11: 74457603-74457603 on Build GRCh38
rs779555364 Chr.11: 74457251-74457251 on Build GRCh38
rs773675377 Chr.11: 74457230-74457230 on Build GRCh38
rs547194943 Chr.11: 74457544-74457544 on Build GRCh38
rs200856070 Chr.11: 74457554-74457554 on Build GRCh38
rs777742722 Chr.11: 74457571-74457571 on Build GRCh38
rs752159282 Chr.11: 74457373-74457373 on Build GRCh38
rs141695803 Chr.11: 74457254-74457254 on Build GRCh38
rs763497447 Chr.11: 74457449-74457450 on Build GRCh38
rs11822977 Chr.11: 74457235-74457235 on Build GRCh38
rs1006962870 Chr.11: 74457290-74457290 on Build GRCh38
rs139040374 Chr.11: 74457538-74457538 on Build GRCh38
rs758367217 Chr.11: 74457382-74457382 on Build GRCh38
rs757172223 Chr.11: 74457451-74457451 on Build GRCh38
rs202036216 Chr.11: 74457349-74457349 on Build GRCh38
rs141880803 Chr.11: 74457444-74457444 on Build GRCh38
rs778413540 Chr.11: 74457369-74457369 on Build GRCh38
rs966495313 Chr.11: 74457317-74457317 on Build GRCh38
rs777664556 Chr.11: 74457383-74457383 on Build GRCh38
rs930193305 Chr.11: 74457516-74457516 on Build GRCh38
rs976125449 Chr.11: 74457308-74457308 on Build GRCh38
rs978419298 Chr.11: 74457322-74457322 on Build GRCh38
rs866853654 Chr.11: 74457415-74457415 on Build GRCh38
rs964790424 Chr.11: 74457306-74457306 on Build GRCh38
rs753448731 Chr.11: 74457257-74457257 on Build GRCh38
rs748088627 Chr.11: 74457269-74457269 on Build GRCh38
rs760938276 Chr.11: 74457342-74457342 on Build GRCh38
rs551016677 Chr.11: 74457620-74457620 on Build GRCh38
rs765329541 Chr.11: 74457562-74457562 on Build GRCh38
rs749496477 Chr.11: 74457399-74457399 on Build GRCh38
rs17215437 Chr.11: 74457316-74457316 on Build GRCh38
rs750315850 Chr.11: 74457593-74457593 on Build GRCh38
rs530975076 Chr.11: 74457427-74457427 on Build GRCh38
rs993767466 Chr.11: 74457231-74457231 on Build GRCh38
rs977986698 Chr.11: 74457605-74457605 on Build GRCh38
rs571006446 Chr.11: 74457650-74457650 on Build GRCh38
rs1009916288 Chr.11: 74457385-74457385 on Build GRCh38
rs775772088 Chr.11: 74457610-74457610 on Build GRCh38
rs542842233 Chr.11: 74457323-74457323 on Build GRCh38
rs748308590 Chr.11: 74457472-74457472 on Build GRCh38
rs747076901 Chr.11: 74457573-74457573 on Build GRCh38
rs762627492 Chr.11: 74457614-74457614 on Build GRCh38
rs565287436 Chr.11: 74457406-74457406 on Build GRCh38
rs775119405 Chr.11: 74457531-74457531 on Build GRCh38
rs868057422 Chr.11: 74457419-74457419 on Build GRCh38
rs759214664 Chr.11: 74457428-74457428 on Build GRCh38
rs368585507 Chr.11: 74457248-74457248 on Build GRCh38
rs557424938 Chr.11: 74457635-74457635 on Build GRCh38
rs201582585 Chr.11: 74457543-74457543 on Build GRCh38
rs746423099 Chr.11: 74457631-74457631 on Build GRCh38
rs375178718 Chr.11: 74457566-74457566 on Build GRCh38
rs200534407 Chr.11: 74457565-74457565 on Build GRCh38
rs762524567 Chr.11: 74457534-74457534 on Build GRCh38
rs121908441 Chr.11: 74457268-74457268 on Build GRCh38
rs17221833 Chr.11: 74457301-74457301 on Build GRCh38
rs772767562 Chr.11: 74457414-74457414 on Build GRCh38
rs771943960 Chr.11: 74457280-74457280 on Build GRCh38
rs199972628 Chr.11: 74457285-74457285 on Build GRCh38
rs530308751 Chr.11: 74457537-74457537 on Build GRCh38
rs545135414 Chr.11: 74457396-74457396 on Build GRCh38
rs755835033 Chr.11: 74457249-74457249 on Build GRCh38
rs888537773 Chr.11: 74457269-74457269 on Build GRCh38
rs754598853 Chr.11: 74457368-74457368 on Build GRCh38
rs2270676 Chr.11: 74457366-74457366 on Build GRCh38
rs767752832 Chr.11: 74457518-74457518 on Build GRCh38
rs766125492 Chr.11: 74457358-74457358 on Build GRCh38
rs768106696 Chr.11: 74457327-74457327 on Build GRCh38
rs17221826 Chr.11: 74457336-74457336 on Build GRCh38
rs929457354 Chr.11: 74457491-74457491 on Build GRCh38
rs557750053 Chr.11: 74457234-74457234 on Build GRCh38
rs764927975 Chr.11: 74457525-74457525 on Build GRCh38
rs755300564 Chr.11: 74457470-74457470 on Build GRCh38
rs547172459 Chr.11: 74457495-74457495 on Build GRCh38
rs375856416 Chr.11: 74457303-74457303 on Build GRCh38
rs752253389 Chr.11: 74457438-74457438 on Build GRCh38
rs768960571 Chr.11: 74457402-74457402 on Build GRCh38
rs556647231 Chr.11: 74457260-74457260 on Build GRCh38
rs761179696 Chr.11: 74457424-74457424 on Build GRCh38
rs17215444 Chr.11: 74457425-74457425 on Build GRCh38
rs773590777 Chr.11: 74457421-74457421 on Build GRCh38
rs34604640 Chr.11: 74457448-74457448 on Build GRCh38
rs764834315 Chr.11: 74457430-74457430 on Build GRCh38
rs371666083 Chr.11: 74457407-74457407 on Build GRCh38

View Full Product Details