Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 66436629-66437130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP2K1
Entrez Gene ID
5604
Gene Name
mitogen-activated protein kinase kinase 1
Gene Aliases
CFC3, MAPKK1, MEK1, MKK1, PRKMK1
UniGene
Hs.145442
Gene Chromosome Location
Chr.15: 66386873-66491544 on Build GRCh38
Transcripts
NM_002755.3 XM_011521783.2 XM_017022411.1 XM_017022412.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3728157 c.308T>A p.I103N Chr.15: 66436762-66436762 on Build GRCh38
1235480 c.387C>A p.F129L Chr.15: 66436841-66436841 on Build GRCh38
7002403 c.293_310del18 p.L98_K104>Q Chr.15: 66436747-66436764 on Build GRCh38
5702512 c.307_312delATCAAA p.I103_K104del Chr.15: 66436761-66436766 on Build GRCh38
4808464 c.389A>G p.Y130C Chr.15: 66436843-66436843 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773454424 Chr.15: 66436725-66436726 on Build GRCh38
rs550240942 Chr.15: 66436900-66436900 on Build GRCh38
rs535697992 Chr.15: 66436925-66436925 on Build GRCh38
rs897186833 Chr.15: 66436886-66436886 on Build GRCh38
rs730880502 Chr.15: 66436762-66436762 on Build GRCh38
rs144166521 Chr.15: 66436769-66436769 on Build GRCh38
rs533695080 Chr.15: 66436881-66436881 on Build GRCh38
rs747647598 Chr.15: 66436697-66436698 on Build GRCh38
rs549156666 Chr.15: 66437050-66437050 on Build GRCh38
rs1039318486 Chr.15: 66437056-66437056 on Build GRCh38
rs749505069 Chr.15: 66436734-66436734 on Build GRCh38
rs913289184 Chr.15: 66437012-66437012 on Build GRCh38
rs397516792 Chr.15: 66436825-66436825 on Build GRCh38
rs750270430 Chr.15: 66436826-66436826 on Build GRCh38
rs797044593 Chr.15: 66436759-66436759 on Build GRCh38
rs375836705 Chr.15: 66437040-66437040 on Build GRCh38
rs939141868 Chr.15: 66436796-66436796 on Build GRCh38
rs753960839 Chr.15: 66436933-66436933 on Build GRCh38
rs1007434866 Chr.15: 66436949-66436949 on Build GRCh38
rs911838368 Chr.15: 66437047-66437047 on Build GRCh38
rs759930943 Chr.15: 66436901-66436901 on Build GRCh38
rs763136654 Chr.15: 66436732-66436732 on Build GRCh38
rs182800877 Chr.15: 66437067-66437067 on Build GRCh38
rs16949944 Chr.15: 66437084-66437084 on Build GRCh38
rs771407622 Chr.15: 66436742-66436742 on Build GRCh38
rs891212917 Chr.15: 66436920-66436920 on Build GRCh38
rs562207701 Chr.15: 66436674-66436674 on Build GRCh38
rs778650731 Chr.15: 66436822-66436822 on Build GRCh38
rs990673168 Chr.15: 66436820-66436820 on Build GRCh38
rs397516793 Chr.15: 66436842-66436842 on Build GRCh38
rs757254963 Chr.15: 66436819-66436819 on Build GRCh38
rs957650379 Chr.15: 66436684-66436684 on Build GRCh38
rs876657651 Chr.15: 66436818-66436818 on Build GRCh38
rs1013906172 Chr.15: 66436833-66436833 on Build GRCh38
rs73471708 Chr.15: 66437095-66437095 on Build GRCh38
rs761016219 Chr.15: 66436930-66436930 on Build GRCh38
rs730880504 Chr.15: 66436866-66436866 on Build GRCh38
rs1018478413 Chr.15: 66437023-66437023 on Build GRCh38
rs773523045 Chr.15: 66436730-66436730 on Build GRCh38
rs934010438 Chr.15: 66437014-66437014 on Build GRCh38
rs369401783 Chr.15: 66436832-66436832 on Build GRCh38
rs527745516 Chr.15: 66436695-66436695 on Build GRCh38
rs1051026685 Chr.15: 66437029-66437029 on Build GRCh38
rs730880508 Chr.15: 66436837-66436838 on Build GRCh38
rs767989355 Chr.15: 66436859-66436859 on Build GRCh38
rs146415623 Chr.15: 66436865-66436865 on Build GRCh38
rs752994941 Chr.15: 66436770-66436770 on Build GRCh38
rs771604017 Chr.15: 66436888-66436888 on Build GRCh38
rs727504819 Chr.15: 66436777-66436777 on Build GRCh38
rs765115125 Chr.15: 66436943-66436943 on Build GRCh38
rs139364105 Chr.15: 66436850-66436850 on Build GRCh38
rs55694358 Chr.15: 66436743-66436743 on Build GRCh38
rs761720615 Chr.15: 66436938-66436938 on Build GRCh38
rs542046069 Chr.15: 66436667-66436667 on Build GRCh38
rs121908595 Chr.15: 66436843-66436843 on Build GRCh38
rs121908596 Chr.15: 66436837-66436837 on Build GRCh38
rs966429123 Chr.15: 66436738-66436738 on Build GRCh38
rs16949939 Chr.15: 66436912-66436912 on Build GRCh38
rs764248591 Chr.15: 66436787-66436787 on Build GRCh38
rs375197930 Chr.15: 66436706-66436706 on Build GRCh38
rs764078823 Chr.15: 66436931-66436931 on Build GRCh38
rs994599054 Chr.15: 66436725-66436725 on Build GRCh38
rs35370745 Chr.15: 66436986-66436987 on Build GRCh38
rs730880503 Chr.15: 66436809-66436809 on Build GRCh38
rs753763967 Chr.15: 66436811-66436811 on Build GRCh38
rs1021954265 Chr.15: 66437089-66437089 on Build GRCh38
rs988917157 Chr.15: 66436951-66436951 on Build GRCh38

View Full Product Details