Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30971027-30971522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHRHR
Entrez Gene ID
2692
Gene Name
growth hormone releasing hormone receptor
Gene Aliases
GHRFR, GRFR, IGHD1B
UniGene
Hs.733003
Gene Chromosome Location
Chr.7: 30964021-30979531 on Build GRCh38
Transcripts
NM_000823.3 XM_011515263.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5438477 c.410A>G p.H137R Chr.7: 30971162-30971162 on Build GRCh38
7241425 c.457G>T p.A153S Chr.7: 30971209-30971209 on Build GRCh38
6514861 c.371C>A p.S124Y Chr.7: 30971123-30971123 on Build GRCh38
6110061 c.460C>G p.L154V Chr.7: 30971212-30971212 on Build GRCh38
3832644 c.417C>G p.I139M Chr.7: 30971169-30971169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs121918118 Chr.7: 30971183-30971183 on Build GRCh38
rs375473377 Chr.7: 30971078-30971078 on Build GRCh38
rs750610125 Chr.7: 30971466-30971466 on Build GRCh38
rs916537711 Chr.7: 30971407-30971407 on Build GRCh38
rs532678098 Chr.7: 30971224-30971224 on Build GRCh38
rs549999412 Chr.7: 30971065-30971065 on Build GRCh38
rs969459740 Chr.7: 30971411-30971411 on Build GRCh38
rs544695401 Chr.7: 30971140-30971140 on Build GRCh38
rs974807573 Chr.7: 30971323-30971323 on Build GRCh38
rs1021667464 Chr.7: 30971154-30971154 on Build GRCh38
rs765003087 Chr.7: 30971151-30971151 on Build GRCh38
rs531764913 Chr.7: 30971060-30971060 on Build GRCh38
rs773214262 Chr.7: 30971109-30971109 on Build GRCh38
rs779187338 Chr.7: 30971210-30971210 on Build GRCh38
rs1053358760 Chr.7: 30971132-30971132 on Build GRCh38
rs781239679 Chr.7: 30971256-30971256 on Build GRCh38
rs764111938 Chr.7: 30971167-30971167 on Build GRCh38
rs900494026 Chr.7: 30971273-30971273 on Build GRCh38
rs574920676 Chr.7: 30971371-30971371 on Build GRCh38
rs973280672 Chr.7: 30971400-30971400 on Build GRCh38
rs923376560 Chr.7: 30971324-30971324 on Build GRCh38
rs947778198 Chr.7: 30971118-30971118 on Build GRCh38
rs1011235926 Chr.7: 30971146-30971146 on Build GRCh38
rs606231412 Chr.7: 30971170-30971170 on Build GRCh38
rs916214234 Chr.7: 30971074-30971074 on Build GRCh38
rs561222414 Chr.7: 30971297-30971297 on Build GRCh38
rs558460416 Chr.7: 30971254-30971254 on Build GRCh38
rs1035420440 Chr.7: 30971169-30971169 on Build GRCh38
rs746126447 Chr.7: 30971263-30971263 on Build GRCh38
rs927670967 Chr.7: 30971422-30971422 on Build GRCh38
rs901989576 Chr.7: 30971064-30971064 on Build GRCh38
rs772602290 Chr.7: 30971087-30971087 on Build GRCh38
rs913400231 Chr.7: 30971421-30971421 on Build GRCh38
rs747453966 Chr.7: 30971247-30971247 on Build GRCh38
rs201267804 Chr.7: 30971155-30971155 on Build GRCh38
rs750156453 Chr.7: 30971200-30971200 on Build GRCh38
rs868202216 Chr.7: 30971175-30971175 on Build GRCh38
rs762844006 Chr.7: 30971159-30971159 on Build GRCh38
rs963207126 Chr.7: 30971369-30971369 on Build GRCh38
rs778375774 Chr.7: 30971245-30971245 on Build GRCh38
rs996230415 Chr.7: 30971283-30971283 on Build GRCh38
rs112879406 Chr.7: 30971124-30971124 on Build GRCh38
rs182348901 Chr.7: 30971238-30971238 on Build GRCh38
rs755821865 Chr.7: 30971203-30971203 on Build GRCh38
rs575933884 Chr.7: 30971272-30971272 on Build GRCh38
rs973013742 Chr.7: 30971211-30971211 on Build GRCh38
rs535557319 Chr.7: 30971173-30971173 on Build GRCh38
rs753197652 Chr.7: 30971199-30971199 on Build GRCh38
rs943372055 Chr.7: 30971286-30971286 on Build GRCh38
rs938811690 Chr.7: 30971144-30971144 on Build GRCh38
rs867907133 Chr.7: 30971165-30971165 on Build GRCh38
rs759699366 Chr.7: 30971147-30971147 on Build GRCh38
rs1001211138 Chr.7: 30971157-30971157 on Build GRCh38
rs573735909 Chr.7: 30971374-30971374 on Build GRCh38
rs1014729472 Chr.7: 30971364-30971364 on Build GRCh38
rs747115030 Chr.7: 30971123-30971123 on Build GRCh38
rs568588560 Chr.7: 30971137-30971137 on Build GRCh38
rs763509630 Chr.7: 30971217-30971217 on Build GRCh38
rs770962593 Chr.7: 30971125-30971125 on Build GRCh38
rs555069923 Chr.7: 30971341-30971341 on Build GRCh38
rs368335119 Chr.7: 30971187-30971187 on Build GRCh38
rs202104123 Chr.7: 30971080-30971080 on Build GRCh38
rs537094779 Chr.7: 30971333-30971333 on Build GRCh38

View Full Product Details