Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100627172-100627662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFR2
Entrez Gene ID
7036
Gene Name
transferrin receptor 2
Gene Aliases
HFE3, TFRC2
UniGene
Hs.544932
Gene Chromosome Location
Chr.7: 100620416-100642780 on Build GRCh38
Transcripts
NM_001206855.1 NM_003227.3 XM_005250553.4 XM_017012573.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4628563 c.1850C>T p.A617V Chr.7: 100627409-100627409 on Build GRCh38
3080551 c.1750G>A p.E584K Chr.7: 100627594-100627594 on Build GRCh38
7422788 c.1967A>T p.N656I Chr.7: 100627292-100627292 on Build GRCh38
3698051 c.1770C>T p.D590D Chr.7: 100627489-100627489 on Build GRCh38
5472303 c.1938C>T p.Y646Y Chr.7: 100627321-100627321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370468237 Chr.7: 100627544-100627544 on Build GRCh38
rs751724123 Chr.7: 100627418-100627418 on Build GRCh38
rs750689567 Chr.7: 100627341-100627341 on Build GRCh38
rs1047318916 Chr.7: 100627533-100627533 on Build GRCh38
rs753158589 Chr.7: 100627369-100627369 on Build GRCh38
rs41295912 Chr.7: 100627570-100627570 on Build GRCh38
rs919600435 Chr.7: 100627514-100627514 on Build GRCh38
rs544210374 Chr.7: 100627467-100627467 on Build GRCh38
rs80338888 Chr.7: 100627387-100627398 on Build GRCh38
rs775586229 Chr.7: 100627643-100627643 on Build GRCh38
rs746098454 Chr.7: 100627632-100627632 on Build GRCh38
rs757861321 Chr.7: 100627510-100627510 on Build GRCh38
rs747923918 Chr.7: 100627537-100627537 on Build GRCh38
rs748879574 Chr.7: 100627273-100627273 on Build GRCh38
rs376552157 Chr.7: 100627471-100627471 on Build GRCh38
rs748426608 Chr.7: 100627228-100627228 on Build GRCh38
rs370288878 Chr.7: 100627381-100627381 on Build GRCh38
rs143251494 Chr.7: 100627582-100627582 on Build GRCh38
rs769498761 Chr.7: 100627243-100627243 on Build GRCh38
rs376149707 Chr.7: 100627379-100627379 on Build GRCh38
rs111760099 Chr.7: 100627318-100627318 on Build GRCh38
rs779551068 Chr.7: 100627511-100627511 on Build GRCh38
rs911003123 Chr.7: 100627495-100627495 on Build GRCh38
rs746499516 Chr.7: 100627588-100627588 on Build GRCh38
rs772314714 Chr.7: 100627642-100627642 on Build GRCh38
rs748036801 Chr.7: 100627602-100627602 on Build GRCh38
rs374393961 Chr.7: 100627545-100627545 on Build GRCh38
rs763294601 Chr.7: 100627413-100627413 on Build GRCh38
rs374489702 Chr.7: 100627396-100627396 on Build GRCh38
rs568798511 Chr.7: 100627302-100627302 on Build GRCh38
rs745441796 Chr.7: 100627506-100627506 on Build GRCh38
rs755908257 Chr.7: 100627253-100627253 on Build GRCh38
rs754267660 Chr.7: 100627564-100627564 on Build GRCh38
rs188214826 Chr.7: 100627518-100627518 on Build GRCh38
rs144723097 Chr.7: 100627619-100627619 on Build GRCh38
rs778041715 Chr.7: 100627375-100627375 on Build GRCh38
rs539909716 Chr.7: 100627339-100627339 on Build GRCh38
rs755561602 Chr.7: 100627365-100627365 on Build GRCh38
rs1012080363 Chr.7: 100627580-100627580 on Build GRCh38
rs770431152 Chr.7: 100627300-100627300 on Build GRCh38
rs368761907 Chr.7: 100627441-100627441 on Build GRCh38
rs1003429018 Chr.7: 100627329-100627329 on Build GRCh38
rs912377680 Chr.7: 100627205-100627205 on Build GRCh38
rs41295915 Chr.7: 100627512-100627512 on Build GRCh38
rs371316105 Chr.7: 100627558-100627558 on Build GRCh38
rs920217192 Chr.7: 100627296-100627296 on Build GRCh38
rs767573164 Chr.7: 100627440-100627440 on Build GRCh38
rs572997617 Chr.7: 100627325-100627325 on Build GRCh38
rs147286030 Chr.7: 100627447-100627447 on Build GRCh38
rs779001760 Chr.7: 100627350-100627350 on Build GRCh38
rs955254550 Chr.7: 100627458-100627458 on Build GRCh38
rs1004353651 Chr.7: 100627411-100627411 on Build GRCh38
rs769477274 Chr.7: 100627604-100627604 on Build GRCh38
rs775278479 Chr.7: 100627395-100627395 on Build GRCh38
rs894600504 Chr.7: 100627572-100627572 on Build GRCh38
rs35704760 Chr.7: 100627489-100627489 on Build GRCh38
rs377756098 Chr.7: 100627549-100627549 on Build GRCh38
rs751548369 Chr.7: 100627364-100627364 on Build GRCh38
rs778534889 Chr.7: 100627384-100627384 on Build GRCh38
rs766491479 Chr.7: 100627414-100627414 on Build GRCh38
rs986685443 Chr.7: 100627195-100627195 on Build GRCh38
rs758031412 Chr.7: 100627581-100627581 on Build GRCh38
rs370375593 Chr.7: 100627597-100627597 on Build GRCh38
rs781639925 Chr.7: 100627214-100627214 on Build GRCh38
rs745338834 Chr.7: 100627390-100627390 on Build GRCh38
rs527901577 Chr.7: 100627551-100627551 on Build GRCh38
rs759516749 Chr.7: 100627436-100627436 on Build GRCh38
rs1056668646 Chr.7: 100627397-100627397 on Build GRCh38
rs377134666 Chr.7: 100627245-100627245 on Build GRCh38
rs781258602 Chr.7: 100627598-100627598 on Build GRCh38
rs560440185 Chr.7: 100627519-100627519 on Build GRCh38
rs2075674 Chr.7: 100627408-100627408 on Build GRCh38
rs374192716 Chr.7: 100627561-100627561 on Build GRCh38
rs775498234 Chr.7: 100627557-100627557 on Build GRCh38
rs1043764677 Chr.7: 100627560-100627560 on Build GRCh38
rs768578503 Chr.7: 100627406-100627406 on Build GRCh38
rs928870996 Chr.7: 100627536-100627536 on Build GRCh38
rs539391212 Chr.7: 100627311-100627311 on Build GRCh38
rs1021635096 Chr.7: 100627407-100627407 on Build GRCh38
rs763205379 Chr.7: 100627349-100627349 on Build GRCh38
rs772114496 Chr.7: 100627391-100627391 on Build GRCh38
rs759540608 Chr.7: 100627320-100627320 on Build GRCh38
rs41295921 Chr.7: 100627255-100627255 on Build GRCh38
rs778131373 Chr.7: 100627451-100627451 on Build GRCh38
rs41295918 Chr.7: 100627357-100627357 on Build GRCh38
rs771543812 Chr.7: 100627410-100627410 on Build GRCh38
rs930230794 Chr.7: 100627304-100627304 on Build GRCh38
rs775396434 Chr.7: 100627335-100627335 on Build GRCh38
rs750051302 Chr.7: 100627571-100627571 on Build GRCh38
rs760383272 Chr.7: 100627336-100627336 on Build GRCh38
rs756482699 Chr.7: 100627370-100627370 on Build GRCh38

View Full Product Details