Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 49200292-49200840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOLH1
Entrez Gene ID
2346
Gene Name
folate hydrolase (prostate-specific membrane antigen) 1
Gene Aliases
FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP
UniGene
Hs.645352
Gene Chromosome Location
Chr.11: 49145094-49208706 on Build GRCh38
Transcripts
NM_001014986.1 NM_001193471.1 NM_001193472.1 NM_001193473.1 NM_004476.1 XM_011519958.2 XM_017017432.1 XM_017017433.1 XM_017017434.1 XM_017017435.1 XM_017017436.1 XM_017017437.1 XM_017017438.1 XM_017017439.1 XM_017017440.1 XM_017017441.1 XM_017017442.1 XM_017017443.1 XM_017017444.1 XM_017017445.1 XM_017017446.1 XM_017017447.1 XM_017017448.1 XM_017017449.1 XM_017017450.1 XM_017017451.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1561379 c.340G>T p.D114Y Chr.11: 49200326-49200326 on Build GRCh38
4821648 c.327G>A p.E109E Chr.11: 49200339-49200339 on Build GRCh38
927691 c.316G>A p.D106N Chr.11: 49200350-49200350 on Build GRCh38
348432 c.289C>G p.Q97E Chr.11: 49200377-49200377 on Build GRCh38
4837709 c.286A>T p.I96F Chr.11: 49200380-49200380 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541224377 Chr.11: 49200665-49200665 on Build GRCh38
rs367766463 Chr.11: 49200425-49200425 on Build GRCh38
rs569807253 Chr.11: 49200534-49200534 on Build GRCh38
rs535760851 Chr.11: 49200655-49200655 on Build GRCh38
rs774791165 Chr.11: 49200422-49200422 on Build GRCh38
rs756233912 Chr.11: 49200457-49200457 on Build GRCh38
rs553946014 Chr.11: 49200727-49200727 on Build GRCh38
rs757517872 Chr.11: 49200477-49200477 on Build GRCh38
rs767202324 Chr.11: 49200744-49200744 on Build GRCh38
rs146289798 Chr.11: 49200339-49200339 on Build GRCh38
rs202680 Chr.11: 49200333-49200333 on Build GRCh38
rs931583108 Chr.11: 49200741-49200741 on Build GRCh38
rs1021651181 Chr.11: 49200585-49200585 on Build GRCh38
rs761483497 Chr.11: 49200390-49200390 on Build GRCh38
rs28759363 Chr.11: 49200557-49200557 on Build GRCh38
rs746506486 Chr.11: 49200366-49200366 on Build GRCh38
rs908869542 Chr.11: 49200775-49200775 on Build GRCh38
rs768200390 Chr.11: 49200367-49200367 on Build GRCh38
rs547139367 Chr.11: 49200342-49200342 on Build GRCh38
rs570533286 Chr.11: 49200353-49200353 on Build GRCh38
rs1011561837 Chr.11: 49200549-49200549 on Build GRCh38
rs975613894 Chr.11: 49200676-49200676 on Build GRCh38
rs754443068 Chr.11: 49200707-49200707 on Build GRCh38
rs745739133 Chr.11: 49200354-49200354 on Build GRCh38
rs752820178 Chr.11: 49200332-49200332 on Build GRCh38
rs768056036 Chr.11: 49200428-49200428 on Build GRCh38
rs185551032 Chr.11: 49200594-49200594 on Build GRCh38
rs555723500 Chr.11: 49200473-49200473 on Build GRCh38
rs772046975 Chr.11: 49200358-49200358 on Build GRCh38
rs753306689 Chr.11: 49200436-49200436 on Build GRCh38
rs897813337 Chr.11: 49200698-49200698 on Build GRCh38
rs766603573 Chr.11: 49200412-49200412 on Build GRCh38
rs1019733156 Chr.11: 49200649-49200649 on Build GRCh38
rs977887921 Chr.11: 49200633-49200633 on Build GRCh38
rs756966079 Chr.11: 49200345-49200345 on Build GRCh38
rs1036805180 Chr.11: 49200424-49200424 on Build GRCh38
rs940400385 Chr.11: 49200786-49200786 on Build GRCh38
rs139621327 Chr.11: 49200362-49200362 on Build GRCh38
rs181091969 Chr.11: 49200573-49200573 on Build GRCh38
rs958615780 Chr.11: 49200540-49200540 on Build GRCh38
rs776233012 Chr.11: 49200389-49200389 on Build GRCh38
rs543024155 Chr.11: 49200766-49200766 on Build GRCh38
rs764303295 Chr.11: 49200469-49200469 on Build GRCh38
rs778510044 Chr.11: 49200488-49200488 on Build GRCh38

View Full Product Details