Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173914463-173914960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINC1
Entrez Gene ID
462
Gene Name
serpin family C member 1
Gene Aliases
AT3, AT3D, ATIII, THPH7
UniGene
Hs.75599
Gene Chromosome Location
Chr.1: 173903804-173917378 on Build GRCh38
Transcripts
NM_000488.3 XM_005245198.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5719413 c.81G>A p.W27* Chr.1: 173914880-173914880 on Build GRCh38
4025227 c.219G>A p.P73P Chr.1: 173914742-173914742 on Build GRCh38
424629 c.147G>A p.M49I Chr.1: 173914814-173914814 on Build GRCh38
899817 c.272C>T p.A91V Chr.1: 173914689-173914689 on Build GRCh38
284908 c.67C>T p.L23F Chr.1: 173914894-173914894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550247582 Chr.1: 173914788-173914788 on Build GRCh38
rs1050200480 Chr.1: 173914752-173914752 on Build GRCh38
rs768704768 Chr.1: 173914828-173914828 on Build GRCh38
rs200810296 Chr.1: 173914610-173914610 on Build GRCh38
rs371222224 Chr.1: 173914598-173914598 on Build GRCh38
rs121909573 Chr.1: 173914582-173914582 on Build GRCh38
rs536867254 Chr.1: 173914525-173914526 on Build GRCh38
rs777405542 Chr.1: 173914794-173914794 on Build GRCh38
rs532883680 Chr.1: 173914873-173914873 on Build GRCh38
rs121909552 Chr.1: 173914725-173914725 on Build GRCh38
rs199469505 Chr.1: 173914750-173914750 on Build GRCh38
rs751112370 Chr.1: 173914883-173914883 on Build GRCh38
rs774294043 Chr.1: 173914728-173914728 on Build GRCh38
rs892676309 Chr.1: 173914835-173914835 on Build GRCh38
rs121909551 Chr.1: 173914743-173914743 on Build GRCh38
rs199469507 Chr.1: 173914725-173914732 on Build GRCh38
rs147266200 Chr.1: 173914696-173914696 on Build GRCh38
rs940626579 Chr.1: 173914700-173914700 on Build GRCh38
rs1002517609 Chr.1: 173914757-173914757 on Build GRCh38
rs745583962 Chr.1: 173914695-173914695 on Build GRCh38
rs377500819 Chr.1: 173914755-173914755 on Build GRCh38
rs948537396 Chr.1: 173914734-173914734 on Build GRCh38
rs892712171 Chr.1: 173914806-173914806 on Build GRCh38
rs531137446 Chr.1: 173914914-173914914 on Build GRCh38
rs121909558 Chr.1: 173914845-173914845 on Build GRCh38
rs147676453 Chr.1: 173914862-173914862 on Build GRCh38
rs121909567 Chr.1: 173914570-173914570 on Build GRCh38
rs775520108 Chr.1: 173914698-173914698 on Build GRCh38
rs28929469 Chr.1: 173914795-173914795 on Build GRCh38
rs369524182 Chr.1: 173914662-173914662 on Build GRCh38
rs754390483 Chr.1: 173914787-173914787 on Build GRCh38
rs765287553 Chr.1: 173914510-173914510 on Build GRCh38
rs145771113 Chr.1: 173914849-173914849 on Build GRCh38
rs374763537 Chr.1: 173914616-173914616 on Build GRCh38
rs758698144 Chr.1: 173914591-173914591 on Build GRCh38
rs387906575 Chr.1: 173914893-173914893 on Build GRCh38
rs374205395 Chr.1: 173914729-173914729 on Build GRCh38
rs369954987 Chr.1: 173914619-173914619 on Build GRCh38
rs878854019 Chr.1: 173914562-173914562 on Build GRCh38
rs764700629 Chr.1: 173914496-173914496 on Build GRCh38
rs747398942 Chr.1: 173914652-173914652 on Build GRCh38
rs773751914 Chr.1: 173914760-173914760 on Build GRCh38
rs201551398 Chr.1: 173914549-173914549 on Build GRCh38
rs1031280047 Chr.1: 173914526-173914526 on Build GRCh38
rs763087077 Chr.1: 173914553-173914553 on Build GRCh38
rs761729771 Chr.1: 173914832-173914832 on Build GRCh38
rs759583638 Chr.1: 173914533-173914533 on Build GRCh38
rs781591817 Chr.1: 173914724-173914724 on Build GRCh38
rs921356636 Chr.1: 173914639-173914639 on Build GRCh38
rs745698642 Chr.1: 173914809-173914809 on Build GRCh38
rs773254902 Chr.1: 173914861-173914861 on Build GRCh38
rs1035248498 Chr.1: 173914532-173914532 on Build GRCh38
rs746429528 Chr.1: 173914742-173914742 on Build GRCh38
rs2227624 Chr.1: 173914872-173914872 on Build GRCh38
rs933978627 Chr.1: 173914656-173914656 on Build GRCh38
rs483352845 Chr.1: 173914931-173914931 on Build GRCh38
rs755827622 Chr.1: 173914929-173914929 on Build GRCh38
rs907768931 Chr.1: 173914677-173914677 on Build GRCh38
rs749308643 Chr.1: 173914827-173914827 on Build GRCh38
rs769761264 Chr.1: 173914819-173914819 on Build GRCh38
rs199895690 Chr.1: 173914659-173914659 on Build GRCh38
rs2227599 Chr.1: 173914937-173914937 on Build GRCh38
rs774506712 Chr.1: 173914829-173914829 on Build GRCh38
rs578203617 Chr.1: 173914548-173914548 on Build GRCh38
rs188274879 Chr.1: 173914874-173914874 on Build GRCh38
rs775397662 Chr.1: 173914823-173914823 on Build GRCh38
rs936295966 Chr.1: 173914768-173914768 on Build GRCh38
rs772807526 Chr.1: 173914507-173914507 on Build GRCh38
rs780733720 Chr.1: 173914911-173914911 on Build GRCh38
rs756940594 Chr.1: 173914799-173914799 on Build GRCh38
rs768646742 Chr.1: 173914936-173914936 on Build GRCh38
rs139392083 Chr.1: 173914611-173914611 on Build GRCh38
rs986804201 Chr.1: 173914915-173914915 on Build GRCh38
rs483352844 Chr.1: 173914838-173914845 on Build GRCh38
rs752544767 Chr.1: 173914778-173914780 on Build GRCh38
rs778100368 Chr.1: 173914646-173914646 on Build GRCh38
rs780670128 Chr.1: 173914801-173914801 on Build GRCh38
rs760408083 Chr.1: 173914509-173914509 on Build GRCh38
rs749037999 Chr.1: 173914934-173914934 on Build GRCh38
rs369828221 Chr.1: 173914902-173914902 on Build GRCh38
rs200118419 Chr.1: 173914650-173914650 on Build GRCh38
rs121909547 Chr.1: 173914726-173914726 on Build GRCh38
rs757851817 Chr.1: 173914647-173914647 on Build GRCh38
rs776257871 Chr.1: 173914511-173914511 on Build GRCh38

View Full Product Details