Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9759252-9759763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR143
Entrez Gene ID
4935
Gene Name
G protein-coupled receptor 143
Gene Aliases
NYS6, OA1
UniGene
Hs.74124
Gene Chromosome Location
Chr.X: 9725413-9765965 on Build GRCh38
Transcripts
NM_000273.2 XM_005274541.3

SNP Detail

SNP ID Chromosome Location
rs373296925 Chr.X: 9759434-9759434 on Build GRCh38
rs779583422 Chr.X: 9759319-9759319 on Build GRCh38
rs1036834522 Chr.X: 9759538-9759538 on Build GRCh38
rs281865180 Chr.X: 9759342-9759343 on Build GRCh38
rs186055724 Chr.X: 9759343-9759343 on Build GRCh38
rs768289150 Chr.X: 9759594-9759594 on Build GRCh38
rs62635762 Chr.X: 9759374-9759374 on Build GRCh38
rs137852296 Chr.X: 9759390-9759390 on Build GRCh38
rs776232889 Chr.X: 9759329-9759329 on Build GRCh38
rs756145241 Chr.X: 9759357-9759357 on Build GRCh38
rs768418077 Chr.X: 9759327-9759327 on Build GRCh38
rs926231983 Chr.X: 9759489-9759489 on Build GRCh38
rs1049086334 Chr.X: 9759629-9759629 on Build GRCh38
rs778099606 Chr.X: 9759372-9759372 on Build GRCh38
rs141573793 Chr.X: 9759424-9759424 on Build GRCh38
rs769585373 Chr.X: 9759477-9759477 on Build GRCh38
rs771403590 Chr.X: 9759497-9759497 on Build GRCh38
rs768027711 Chr.X: 9759439-9759439 on Build GRCh38
rs748188612 Chr.X: 9759325-9759325 on Build GRCh38
rs58068096 Chr.X: 9759347-9759347 on Build GRCh38
rs930652482 Chr.X: 9759565-9759565 on Build GRCh38
rs917373425 Chr.X: 9759524-9759524 on Build GRCh38
rs776290292 Chr.X: 9759455-9759455 on Build GRCh38
rs281865181 Chr.X: 9759331-9759331 on Build GRCh38
rs757567458 Chr.X: 9759472-9759472 on Build GRCh38
rs62635032 Chr.X: 9759416-9759416 on Build GRCh38
rs780741308 Chr.X: 9759710-9759710 on Build GRCh38
rs973943974 Chr.X: 9759508-9759508 on Build GRCh38
rs557790936 Chr.X: 9759664-9759664 on Build GRCh38
rs769824928 Chr.X: 9759459-9759459 on Build GRCh38
rs746336091 Chr.X: 9759322-9759322 on Build GRCh38
rs746567246 Chr.X: 9759435-9759435 on Build GRCh38
rs761647216 Chr.X: 9759340-9759340 on Build GRCh38
rs748890250 Chr.X: 9759578-9759578 on Build GRCh38
rs773891058 Chr.X: 9759605-9759605 on Build GRCh38
rs771486573 Chr.X: 9759323-9759323 on Build GRCh38
rs62635034 Chr.X: 9759385-9759385 on Build GRCh38
rs767508273 Chr.X: 9759350-9759350 on Build GRCh38
rs761262655 Chr.X: 9759708-9759708 on Build GRCh38
rs754062843 Chr.X: 9759376-9759376 on Build GRCh38
rs777116393 Chr.X: 9759536-9759536 on Build GRCh38
rs58933950 Chr.X: 9759332-9759332 on Build GRCh38
rs62635762 Chr.X: 9759374-9759374 on Build GRCh38
rs942749507 Chr.X: 9759546-9759546 on Build GRCh38
rs772688504 Chr.X: 9759426-9759426 on Build GRCh38
rs912633071 Chr.X: 9759540-9759540 on Build GRCh38
rs762815553 Chr.X: 9759344-9759344 on Build GRCh38
rs749739235 Chr.X: 9759300-9759300 on Build GRCh38
rs35528779 Chr.X: 9759387-9759388 on Build GRCh38
rs918388063 Chr.X: 9759521-9759521 on Build GRCh38
rs758911399 Chr.X: 9759552-9759552 on Build GRCh38
rs930078612 Chr.X: 9759535-9759535 on Build GRCh38
rs181078528 Chr.X: 9759312-9759312 on Build GRCh38
rs148358078 Chr.X: 9759391-9759391 on Build GRCh38
rs937593614 Chr.X: 9759496-9759496 on Build GRCh38
rs201830411 Chr.X: 9759351-9759351 on Build GRCh38

View Full Product Details