Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 23919817-23920387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL15
Entrez Gene ID
6138
Gene Name
ribosomal protein L15
Gene Aliases
DBA12, EC45, L15, RPL10, RPLY10, RPYL10
UniGene
Hs.381219
Gene Chromosome Location
Chr.3: 23916804-23923696 on Build GRCh38
Transcripts
NM_001253379.1 NM_001253380.1 NM_001253382.1 NM_001253383.1 NM_001253384.1 NM_002948.3

SNP Detail

SNP ID Chromosome Location
rs780330382 Chr.3: 23920313-23920313 on Build GRCh38
rs867732845 Chr.3: 23920281-23920281 on Build GRCh38
rs1030701654 Chr.3: 23919892-23919892 on Build GRCh38
rs779286291 Chr.3: 23920262-23920262 on Build GRCh38
rs927843711 Chr.3: 23920284-23920284 on Build GRCh38
rs537698656 Chr.3: 23920287-23920287 on Build GRCh38
rs752177706 Chr.3: 23919987-23919987 on Build GRCh38
rs955108818 Chr.3: 23920204-23920204 on Build GRCh38
rs558835275 Chr.3: 23920296-23920296 on Build GRCh38
rs1045353512 Chr.3: 23919881-23919881 on Build GRCh38
rs569286254 Chr.3: 23920178-23920187 on Build GRCh38
rs535298390 Chr.3: 23920329-23920329 on Build GRCh38
rs564924527 Chr.3: 23919891-23919892 on Build GRCh38
rs34082882 Chr.3: 23919862-23919862 on Build GRCh38
rs1044946432 Chr.3: 23920080-23920080 on Build GRCh38
rs1133926 Chr.3: 23920271-23920271 on Build GRCh38
rs535144479 Chr.3: 23919839-23919839 on Build GRCh38
rs182309439 Chr.3: 23919890-23919890 on Build GRCh38
rs116295721 Chr.3: 23920093-23920093 on Build GRCh38
rs57810037 Chr.3: 23919847-23919847 on Build GRCh38
rs373934726 Chr.3: 23919918-23919919 on Build GRCh38
rs535888300 Chr.3: 23919871-23919871 on Build GRCh38
rs899567598 Chr.3: 23920363-23920363 on Build GRCh38
rs34274724 Chr.3: 23920075-23920076 on Build GRCh38
rs1052323478 Chr.3: 23920300-23920300 on Build GRCh38
rs1052514182 Chr.3: 23920094-23920094 on Build GRCh38
rs758942175 Chr.3: 23920305-23920305 on Build GRCh38
rs559742573 Chr.3: 23919893-23919893 on Build GRCh38
rs961987531 Chr.3: 23920222-23920222 on Build GRCh38
rs542313336 Chr.3: 23919962-23919962 on Build GRCh38
rs11129128 Chr.3: 23920130-23920130 on Build GRCh38
rs973428228 Chr.3: 23920068-23920068 on Build GRCh38
rs748550537 Chr.3: 23920316-23920318 on Build GRCh38
rs558283953 Chr.3: 23919966-23919966 on Build GRCh38
rs899622964 Chr.3: 23920165-23920165 on Build GRCh38
rs371102571 Chr.3: 23920104-23920104 on Build GRCh38
rs567690775 Chr.3: 23920176-23920176 on Build GRCh38
rs755818088 Chr.3: 23920038-23920038 on Build GRCh38
rs974941036 Chr.3: 23920003-23920003 on Build GRCh38
rs140106884 Chr.3: 23919848-23919848 on Build GRCh38
rs780453851 Chr.3: 23920366-23920366 on Build GRCh38
rs906436203 Chr.3: 23920089-23920089 on Build GRCh38
rs574999516 Chr.3: 23919917-23919917 on Build GRCh38
rs142246641 Chr.3: 23920109-23920109 on Build GRCh38
rs537730308 Chr.3: 23920218-23920218 on Build GRCh38
rs367621594 Chr.3: 23919899-23919900 on Build GRCh38
rs751925811 Chr.3: 23920036-23920036 on Build GRCh38
rs753460903 Chr.3: 23920102-23920102 on Build GRCh38
rs973380922 Chr.3: 23920261-23920261 on Build GRCh38
rs968839297 Chr.3: 23919929-23919929 on Build GRCh38
rs1029655453 Chr.3: 23920013-23920013 on Build GRCh38
rs546205454 Chr.3: 23920158-23920158 on Build GRCh38
rs1141621 Chr.3: 23920367-23920367 on Build GRCh38
rs755225847 Chr.3: 23920262-23920264 on Build GRCh38
rs913822613 Chr.3: 23920301-23920301 on Build GRCh38
rs185297056 Chr.3: 23919854-23919854 on Build GRCh38
rs1022071974 Chr.3: 23919925-23919925 on Build GRCh38
rs947843440 Chr.3: 23920075-23920075 on Build GRCh38
rs191714155 Chr.3: 23920354-23920354 on Build GRCh38
rs987862953 Chr.3: 23920039-23920039 on Build GRCh38
rs757361112 Chr.3: 23919844-23919844 on Build GRCh38
rs571131453 Chr.3: 23920311-23920311 on Build GRCh38
rs535061537 Chr.3: 23920201-23920201 on Build GRCh38
rs536813835 Chr.3: 23920294-23920294 on Build GRCh38
rs79806018 Chr.3: 23920064-23920064 on Build GRCh38
rs907903453 Chr.3: 23920050-23920050 on Build GRCh38
rs187233990 Chr.3: 23920026-23920026 on Build GRCh38
rs898977322 Chr.3: 23919855-23919855 on Build GRCh38
rs189958810 Chr.3: 23919866-23919866 on Build GRCh38
rs148622248 Chr.3: 23920216-23920216 on Build GRCh38

View Full Product Details