Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27463249-27463832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA2
Entrez Gene ID
1135
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Gene Aliases
-
UniGene
Hs.57718
Gene Chromosome Location
Chr.8: 27459761-27479296 on Build GRCh38
Transcripts
NM_000742.3 NM_001282455.1 XM_005273397.2 XM_006716282.1 XM_011544388.1 XM_011544389.1 XM_017013003.1 XM_017013004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7157974 c.1148C>G p.P383R Chr.8: 27463295-27463295 on Build GRCh38
1201117 c.719A>G p.Y240C Chr.8: 27463724-27463724 on Build GRCh38
5976802 c.697A>C p.I233L Chr.8: 27463746-27463746 on Build GRCh38
5034909 c.861C>T p.F287F Chr.8: 27463582-27463582 on Build GRCh38
5874476 c.1103G>A p.G368E Chr.8: 27463340-27463340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756903177 Chr.8: 27463648-27463648 on Build GRCh38
rs546670055 Chr.8: 27463799-27463799 on Build GRCh38
rs762812430 Chr.8: 27463490-27463490 on Build GRCh38
rs139727115 Chr.8: 27463560-27463560 on Build GRCh38
rs929539214 Chr.8: 27463689-27463689 on Build GRCh38
rs56229264 Chr.8: 27463672-27463672 on Build GRCh38
rs748234276 Chr.8: 27463784-27463784 on Build GRCh38
rs768590131 Chr.8: 27463742-27463742 on Build GRCh38
rs758403895 Chr.8: 27463376-27463376 on Build GRCh38
rs143360318 Chr.8: 27463450-27463450 on Build GRCh38
rs777491869 Chr.8: 27463533-27463533 on Build GRCh38
rs560790452 Chr.8: 27463285-27463285 on Build GRCh38
rs374512926 Chr.8: 27463316-27463316 on Build GRCh38
rs886042322 Chr.8: 27463506-27463506 on Build GRCh38
rs768696312 Chr.8: 27463555-27463555 on Build GRCh38
rs114294066 Chr.8: 27463530-27463530 on Build GRCh38
rs758255768 Chr.8: 27463424-27463424 on Build GRCh38
rs56339365 Chr.8: 27463380-27463380 on Build GRCh38
rs587780891 Chr.8: 27463498-27463498 on Build GRCh38
rs796052309 Chr.8: 27463554-27463556 on Build GRCh38
rs998048218 Chr.8: 27463586-27463586 on Build GRCh38
rs544814869 Chr.8: 27463719-27463719 on Build GRCh38
rs749494357 Chr.8: 27463284-27463284 on Build GRCh38
rs151068685 Chr.8: 27463537-27463537 on Build GRCh38
rs1050947622 Chr.8: 27463756-27463756 on Build GRCh38
rs767782606 Chr.8: 27463345-27463345 on Build GRCh38
rs79996627 Chr.8: 27463446-27463446 on Build GRCh38
rs908715626 Chr.8: 27463525-27463525 on Build GRCh38
rs767656036 Chr.8: 27463406-27463406 on Build GRCh38
rs767492985 Chr.8: 27463707-27463707 on Build GRCh38
rs933808461 Chr.8: 27463748-27463748 on Build GRCh38
rs1009284130 Chr.8: 27463444-27463444 on Build GRCh38
rs776429147 Chr.8: 27463383-27463383 on Build GRCh38
rs796052300 Chr.8: 27463718-27463718 on Build GRCh38
rs752194489 Chr.8: 27463579-27463579 on Build GRCh38
rs768424025 Chr.8: 27463325-27463325 on Build GRCh38
rs746292157 Chr.8: 27463653-27463653 on Build GRCh38
rs149774326 Chr.8: 27463629-27463629 on Build GRCh38
rs746613509 Chr.8: 27463320-27463320 on Build GRCh38
rs796052303 Chr.8: 27463278-27463278 on Build GRCh38
rs750130500 Chr.8: 27463487-27463487 on Build GRCh38
rs756924174 Chr.8: 27463727-27463727 on Build GRCh38
rs201370407 Chr.8: 27463680-27463680 on Build GRCh38
rs146751925 Chr.8: 27463712-27463712 on Build GRCh38
rs143536618 Chr.8: 27463759-27463759 on Build GRCh38
rs762608232 Chr.8: 27463561-27463561 on Build GRCh38
rs1002855646 Chr.8: 27463464-27463464 on Build GRCh38
rs760884840 Chr.8: 27463484-27463484 on Build GRCh38
rs770305730 Chr.8: 27463391-27463391 on Build GRCh38
rs202127295 Chr.8: 27463577-27463577 on Build GRCh38
rs188098799 Chr.8: 27463299-27463299 on Build GRCh38
rs567044128 Chr.8: 27463413-27463413 on Build GRCh38
rs750472173 Chr.8: 27463617-27463618 on Build GRCh38
rs201279312 Chr.8: 27463791-27463791 on Build GRCh38
rs104894063 Chr.8: 27463607-27463607 on Build GRCh38
rs747493891 Chr.8: 27463755-27463755 on Build GRCh38
rs771531957 Chr.8: 27463396-27463396 on Build GRCh38
rs751699392 Chr.8: 27463344-27463344 on Build GRCh38
rs144913811 Chr.8: 27463684-27463684 on Build GRCh38
rs762406891 Chr.8: 27463269-27463269 on Build GRCh38
rs368791756 Chr.8: 27463379-27463379 on Build GRCh38
rs753568467 Chr.8: 27463348-27463348 on Build GRCh38
rs779197511 Chr.8: 27463543-27463543 on Build GRCh38
rs554976506 Chr.8: 27463513-27463513 on Build GRCh38
rs757169328 Chr.8: 27463296-27463296 on Build GRCh38
rs745774113 Chr.8: 27463375-27463375 on Build GRCh38
rs750301958 Chr.8: 27463422-27463422 on Build GRCh38
rs376807512 Chr.8: 27463656-27463656 on Build GRCh38
rs374766524 Chr.8: 27463588-27463588 on Build GRCh38
rs74341575 Chr.8: 27463370-27463370 on Build GRCh38
rs775595773 Chr.8: 27463708-27463708 on Build GRCh38
rs763633128 Chr.8: 27463714-27463714 on Build GRCh38
rs1040149262 Chr.8: 27463696-27463696 on Build GRCh38
rs780941610 Chr.8: 27463441-27463441 on Build GRCh38
rs780056270 Chr.8: 27463429-27463429 on Build GRCh38
rs375239013 Chr.8: 27463486-27463486 on Build GRCh38
rs778777217 Chr.8: 27463372-27463372 on Build GRCh38
rs769608818 Chr.8: 27463453-27463453 on Build GRCh38
rs773886433 Chr.8: 27463342-27463342 on Build GRCh38
rs796052302 Chr.8: 27463295-27463295 on Build GRCh38
rs751751026 Chr.8: 27463289-27463289 on Build GRCh38
rs886062844 Chr.8: 27463280-27463280 on Build GRCh38
rs760940111 Chr.8: 27463414-27463414 on Build GRCh38
rs754567543 Chr.8: 27463436-27463436 on Build GRCh38
rs892421066 Chr.8: 27463633-27463633 on Build GRCh38
rs781326114 Chr.8: 27463773-27463773 on Build GRCh38
rs775592017 Chr.8: 27463478-27463478 on Build GRCh38
rs367914235 Chr.8: 27463538-27463538 on Build GRCh38
rs968471265 Chr.8: 27463658-27463658 on Build GRCh38
rs748482434 Chr.8: 27463384-27463384 on Build GRCh38
rs56371471 Chr.8: 27463458-27463458 on Build GRCh38
rs371858399 Chr.8: 27463459-27463459 on Build GRCh38
rs759490278 Chr.8: 27463463-27463463 on Build GRCh38
rs772635760 Chr.8: 27463550-27463550 on Build GRCh38
rs77769511 Chr.8: 27463666-27463666 on Build GRCh38
rs762282272 Chr.8: 27463338-27463338 on Build GRCh38
rs147530139 Chr.8: 27463456-27463456 on Build GRCh38
rs758848678 Chr.8: 27463654-27463654 on Build GRCh38
rs1039458194 Chr.8: 27463732-27463732 on Build GRCh38
rs755274565 Chr.8: 27463290-27463290 on Build GRCh38
rs140118184 Chr.8: 27463477-27463477 on Build GRCh38
rs77710085 Chr.8: 27463698-27463698 on Build GRCh38
rs888244000 Chr.8: 27463298-27463298 on Build GRCh38
rs775537123 Chr.8: 27463551-27463551 on Build GRCh38
rs775471233 Chr.8: 27463807-27463807 on Build GRCh38
rs771282047 Chr.8: 27463457-27463457 on Build GRCh38
rs1039488837 Chr.8: 27463563-27463563 on Build GRCh38
rs763337219 Chr.8: 27463393-27463393 on Build GRCh38
rs137965290 Chr.8: 27463326-27463326 on Build GRCh38
rs554981272 Chr.8: 27463564-27463564 on Build GRCh38
rs372418511 Chr.8: 27463683-27463683 on Build GRCh38
rs542738759 Chr.8: 27463268-27463268 on Build GRCh38
rs796052301 Chr.8: 27463359-27463359 on Build GRCh38
rs769975305 Chr.8: 27463681-27463681 on Build GRCh38
rs879129089 Chr.8: 27463747-27463747 on Build GRCh38
rs760660239 Chr.8: 27463809-27463809 on Build GRCh38
rs74929100 Chr.8: 27463401-27463401 on Build GRCh38
rs765772636 Chr.8: 27463340-27463340 on Build GRCh38
rs774054592 Chr.8: 27463569-27463569 on Build GRCh38
rs774279507 Chr.8: 27463398-27463398 on Build GRCh38
rs767685409 Chr.8: 27463473-27463473 on Build GRCh38
rs200372256 Chr.8: 27463671-27463671 on Build GRCh38
rs56189058 Chr.8: 27463609-27463609 on Build GRCh38
rs200523775 Chr.8: 27463792-27463792 on Build GRCh38
rs1009464639 Chr.8: 27463808-27463808 on Build GRCh38
rs183676062 Chr.8: 27463270-27463270 on Build GRCh38
rs758793756 Chr.8: 27463743-27463743 on Build GRCh38
rs140350483 Chr.8: 27463733-27463733 on Build GRCh38
rs757376257 Chr.8: 27463337-27463338 on Build GRCh38
rs777510962 Chr.8: 27463292-27463292 on Build GRCh38
rs766188212 Chr.8: 27463504-27463504 on Build GRCh38
rs745346653 Chr.8: 27463301-27463301 on Build GRCh38
rs767619637 Chr.8: 27463574-27463574 on Build GRCh38
rs771508746 Chr.8: 27463304-27463304 on Build GRCh38
rs529695608 Chr.8: 27463796-27463796 on Build GRCh38
rs148608618 Chr.8: 27463449-27463449 on Build GRCh38
rs1018084204 Chr.8: 27463317-27463317 on Build GRCh38
rs747107008 Chr.8: 27463553-27463553 on Build GRCh38
rs773759759 Chr.8: 27463392-27463392 on Build GRCh38
rs867857056 Chr.8: 27463353-27463353 on Build GRCh38
rs780491416 Chr.8: 27463746-27463746 on Build GRCh38
rs149464248 Chr.8: 27463343-27463343 on Build GRCh38
rs746347227 Chr.8: 27463547-27463547 on Build GRCh38
rs369173344 Chr.8: 27463597-27463597 on Build GRCh38
rs752321195 Chr.8: 27463519-27463519 on Build GRCh38
rs888935641 Chr.8: 27463367-27463367 on Build GRCh38
rs755014350 Chr.8: 27463518-27463518 on Build GRCh38
rs992286474 Chr.8: 27463501-27463501 on Build GRCh38
rs377660408 Chr.8: 27463790-27463790 on Build GRCh38
rs76140563 Chr.8: 27463570-27463570 on Build GRCh38
rs749144601 Chr.8: 27463294-27463294 on Build GRCh38
rs775230859 Chr.8: 27463455-27463455 on Build GRCh38
rs145730990 Chr.8: 27463655-27463655 on Build GRCh38

View Full Product Details