Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 40798584-40799177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A15
Entrez Gene ID
10166
Gene Name
solute carrier family 25 member 15
Gene Aliases
D13S327, HHH, ORC1, ORNT1
UniGene
Hs.646645
Gene Chromosome Location
Chr.13: 40789411-40812460 on Build GRCh38
Transcripts
NM_014252.3 NR_038258.1 NR_038259.1

Gene Symbol
TPTE2P5
Entrez Gene ID
100616668
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 5
Gene Aliases
-
UniGene
Hs.660718
Gene Chromosome Location
Chr.13: 40796985-40921750 on Build GRCh38
Transcripts
NM_014252.3 NR_038258.1 NR_038259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4726791 c.121C>A p.P41T Chr.13: 40799122-40799122 on Build GRCh38
432391 c.159G>T p.K53N Chr.13: 40799160-40799160 on Build GRCh38
3738436 c.84G>T p.Q28H Chr.13: 40799085-40799085 on Build GRCh38
6074685 c.91G>C p.D31H Chr.13: 40799092-40799092 on Build GRCh38
1747440 c.56G>A p.G19E Chr.13: 40799057-40799057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529807706 Chr.13: 40798746-40798746 on Build GRCh38
rs201725044 Chr.13: 40799026-40799026 on Build GRCh38
rs776411120 Chr.13: 40799089-40799089 on Build GRCh38
rs567609947 Chr.13: 40798996-40798996 on Build GRCh38
rs929319558 Chr.13: 40798976-40798976 on Build GRCh38
rs987719565 Chr.13: 40798825-40798825 on Build GRCh38
rs781334378 Chr.13: 40798884-40798884 on Build GRCh38
rs760167822 Chr.13: 40799071-40799071 on Build GRCh38
rs764024928 Chr.13: 40799117-40799117 on Build GRCh38
rs144656633 Chr.13: 40799140-40799140 on Build GRCh38
rs1044512136 Chr.13: 40798792-40798792 on Build GRCh38
rs201543522 Chr.13: 40799031-40799031 on Build GRCh38
rs752415542 Chr.13: 40799097-40799100 on Build GRCh38
rs2297013 Chr.13: 40798808-40798808 on Build GRCh38
rs752660500 Chr.13: 40799017-40799017 on Build GRCh38
rs781152683 Chr.13: 40799029-40799029 on Build GRCh38
rs372006813 Chr.13: 40799051-40799051 on Build GRCh38
rs121908536 Chr.13: 40799096-40799096 on Build GRCh38
rs752490809 Chr.13: 40799094-40799094 on Build GRCh38
rs540292884 Chr.13: 40799134-40799134 on Build GRCh38
rs370453495 Chr.13: 40798862-40798862 on Build GRCh38
rs147032124 Chr.13: 40798809-40798809 on Build GRCh38
rs939873464 Chr.13: 40798698-40798698 on Build GRCh38
rs2297014 Chr.13: 40798824-40798824 on Build GRCh38
rs368677375 Chr.13: 40799044-40799044 on Build GRCh38
rs776040168 Chr.13: 40798699-40798699 on Build GRCh38
rs961271480 Chr.13: 40798982-40798982 on Build GRCh38
rs759403059 Chr.13: 40799006-40799006 on Build GRCh38
rs931090205 Chr.13: 40798761-40798761 on Build GRCh38
rs758838019 Chr.13: 40799150-40799150 on Build GRCh38
rs759454598 Chr.13: 40799092-40799092 on Build GRCh38
rs193114015 Chr.13: 40798752-40798752 on Build GRCh38
rs71427425 Chr.13: 40798657-40798657 on Build GRCh38
rs972616130 Chr.13: 40798991-40798991 on Build GRCh38
rs762809317 Chr.13: 40799101-40799101 on Build GRCh38
rs41396747 Chr.13: 40799118-40799118 on Build GRCh38
rs987202579 Chr.13: 40798967-40798967 on Build GRCh38
rs756292519 Chr.13: 40799040-40799040 on Build GRCh38
rs746014793 Chr.13: 40798886-40798886 on Build GRCh38
rs973485322 Chr.13: 40798939-40798939 on Build GRCh38
rs756522093 Chr.13: 40799112-40799113 on Build GRCh38
rs552144413 Chr.13: 40798692-40798692 on Build GRCh38
rs778342930 Chr.13: 40799153-40799153 on Build GRCh38
rs920585443 Chr.13: 40798958-40798958 on Build GRCh38
rs545685860 Chr.13: 40798969-40798969 on Build GRCh38
rs104894430 Chr.13: 40799080-40799080 on Build GRCh38
rs1032008747 Chr.13: 40798769-40798769 on Build GRCh38
rs57687350 Chr.13: 40798822-40798823 on Build GRCh38
rs762979476 Chr.13: 40798786-40798786 on Build GRCh38
rs779433260 Chr.13: 40799048-40799048 on Build GRCh38
rs201647638 Chr.13: 40799135-40799135 on Build GRCh38
rs376634191 Chr.13: 40799145-40799145 on Build GRCh38
rs1047942272 Chr.13: 40798979-40798979 on Build GRCh38
rs954144556 Chr.13: 40798668-40798668 on Build GRCh38
rs770041563 Chr.13: 40799004-40799004 on Build GRCh38
rs1024685863 Chr.13: 40798725-40798725 on Build GRCh38
rs780317136 Chr.13: 40799042-40799042 on Build GRCh38
rs187685447 Chr.13: 40799148-40799148 on Build GRCh38
rs904948403 Chr.13: 40798735-40798735 on Build GRCh38
rs121908533 Chr.13: 40799111-40799111 on Build GRCh38
rs546842193 Chr.13: 40798606-40798606 on Build GRCh38
rs560300254 Chr.13: 40799139-40799139 on Build GRCh38
rs538277665 Chr.13: 40798832-40798832 on Build GRCh38
rs772613660 Chr.13: 40799069-40799069 on Build GRCh38
rs767135599 Chr.13: 40799058-40799063 on Build GRCh38
rs367928183 Chr.13: 40799013-40799013 on Build GRCh38
rs199737843 Chr.13: 40799099-40799099 on Build GRCh38
rs1047992807 Chr.13: 40798681-40798681 on Build GRCh38
rs943351539 Chr.13: 40798630-40798630 on Build GRCh38
rs539373322 Chr.13: 40799146-40799146 on Build GRCh38
rs17061572 Chr.13: 40798675-40798675 on Build GRCh38
rs896724945 Chr.13: 40798823-40798823 on Build GRCh38
rs1001934900 Chr.13: 40799041-40799041 on Build GRCh38
rs979881918 Chr.13: 40799025-40799025 on Build GRCh38
rs1014853139 Chr.13: 40798722-40798722 on Build GRCh38
rs573826314 Chr.13: 40798816-40798816 on Build GRCh38
rs369247293 Chr.13: 40798631-40798631 on Build GRCh38
rs903996426 Chr.13: 40798861-40798861 on Build GRCh38
rs772586133 Chr.13: 40799065-40799065 on Build GRCh38
rs573099150 Chr.13: 40799041-40799042 on Build GRCh38
rs753050575 Chr.13: 40798667-40798667 on Build GRCh38

View Full Product Details