Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38414163-38414335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFR1
Entrez Gene ID
2260
Gene Name
fibroblast growth factor receptor 1
Gene Aliases
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
UniGene
Hs.264887
Gene Chromosome Location
Chr.8: 38411138-38468834 on Build GRCh38
Transcripts
NM_001174063.1 NM_001174064.1 NM_001174065.1 NM_001174066.1 NM_001174067.1 NM_015850.3 NM_023105.2 NM_023106.2 NM_023110.2 XM_006716303.2 XM_006716304.1 XM_006716306.2 XM_006716307.1 XM_006716309.3 XM_006716310.2 XM_006716311.1 XM_006716312.1 XM_006716313.2 XM_006716314.1 XM_011544443.2 XM_011544444.1 XM_011544445.2 XM_011544446.2 XM_011544447.2 XM_011544448.1 XM_011544449.1 XM_011544450.2 XM_011544451.1 XM_017013219.1 XM_017013220.1 XM_017013221.1 XM_017013222.1 XM_017013223.1 XM_017013224.1 XM_017013225.1 XM_017013226.1 XM_017013227.1 XM_017013228.1 XM_017013229.1 XM_017013230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5781110 c.2119G>C p.E707Q Chr.8: 38414219-38414219 on Build GRCh38
3395314 c.2092G>A p.G698S Chr.8: 38414246-38414246 on Build GRCh38
6933851 c.2152C>A p.R718S Chr.8: 38414186-38414186 on Build GRCh38
3834665 c.2050T>G p.W684G Chr.8: 38414288-38414288 on Build GRCh38
311185 c.2091C>A p.G697G Chr.8: 38414247-38414247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768957161 Chr.8: 38414231-38414231 on Build GRCh38
rs780575123 Chr.8: 38414211-38414211 on Build GRCh38
rs999999057 Chr.8: 38414315-38414315 on Build GRCh38
rs763166714 Chr.8: 38414261-38414261 on Build GRCh38
rs757055030 Chr.8: 38414294-38414294 on Build GRCh38
rs515726225 Chr.8: 38414254-38414254 on Build GRCh38
rs748318003 Chr.8: 38414241-38414241 on Build GRCh38
rs766598761 Chr.8: 38414271-38414271 on Build GRCh38
rs758877273 Chr.8: 38414208-38414208 on Build GRCh38
rs952285515 Chr.8: 38414268-38414268 on Build GRCh38
rs760700056 Chr.8: 38414298-38414298 on Build GRCh38
rs747542969 Chr.8: 38414217-38414217 on Build GRCh38
rs397515445 Chr.8: 38414263-38414263 on Build GRCh38
rs756845879 Chr.8: 38414302-38414302 on Build GRCh38
rs777061347 Chr.8: 38414232-38414232 on Build GRCh38
rs1057518060 Chr.8: 38414276-38414276 on Build GRCh38
rs727505376 Chr.8: 38414279-38414279 on Build GRCh38
rs745441616 Chr.8: 38414197-38414197 on Build GRCh38
rs775520462 Chr.8: 38414294-38414294 on Build GRCh38
rs778281261 Chr.8: 38414193-38414193 on Build GRCh38
rs773144331 Chr.8: 38414247-38414247 on Build GRCh38
rs563601371 Chr.8: 38414301-38414301 on Build GRCh38
rs770115570 Chr.8: 38414244-38414244 on Build GRCh38
rs876661335 Chr.8: 38414264-38414264 on Build GRCh38
rs370044394 Chr.8: 38414305-38414305 on Build GRCh38

View Full Product Details