Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38415824-38415985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGFR1
Entrez Gene ID
2260
Gene Name
fibroblast growth factor receptor 1
Gene Aliases
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1
UniGene
Hs.264887
Gene Chromosome Location
Chr.8: 38411138-38468834 on Build GRCh38
Transcripts
NM_001174063.1 NM_001174064.1 NM_001174065.1 NM_001174066.1 NM_001174067.1 NM_015850.3 NM_023105.2 NM_023106.2 NM_023110.2 XM_006716303.2 XM_006716304.1 XM_006716306.2 XM_006716307.1 XM_006716309.3 XM_006716310.2 XM_006716311.1 XM_006716312.1 XM_006716313.2 XM_006716314.1 XM_011544443.2 XM_011544444.1 XM_011544445.2 XM_011544446.2 XM_011544447.2 XM_011544448.1 XM_011544449.1 XM_011544450.2 XM_011544451.1 XM_017013219.1 XM_017013220.1 XM_017013221.1 XM_017013222.1 XM_017013223.1 XM_017013224.1 XM_017013225.1 XM_017013226.1 XM_017013227.1 XM_017013228.1 XM_017013229.1 XM_017013230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2960434 c.1760C>T p.S587F Chr.8: 38415934-38415934 on Build GRCh38
1597691 c.1779C>T p.C593C Chr.8: 38415915-38415915 on Build GRCh38
4684544 c.1772C>A p.P591Q Chr.8: 38415952-38415952 on Build GRCh38
6912692 c.1823C>A p.A608D Chr.8: 38415901-38415901 on Build GRCh38
4367394 c.1809C>T p.C603C Chr.8: 38415915-38415915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770658262 Chr.8: 38415850-38415850 on Build GRCh38
rs199830036 Chr.8: 38415851-38415851 on Build GRCh38
rs758626967 Chr.8: 38415912-38415912 on Build GRCh38
rs768986378 Chr.8: 38415879-38415879 on Build GRCh38
rs926779719 Chr.8: 38415948-38415948 on Build GRCh38
rs1057518218 Chr.8: 38415950-38415960 on Build GRCh38
rs765629435 Chr.8: 38415898-38415898 on Build GRCh38
rs771678301 Chr.8: 38415861-38415861 on Build GRCh38
rs371019935 Chr.8: 38415888-38415888 on Build GRCh38
rs777228638 Chr.8: 38415881-38415881 on Build GRCh38
rs755002934 Chr.8: 38415950-38415950 on Build GRCh38
rs866906610 Chr.8: 38415926-38415926 on Build GRCh38
rs755580322 Chr.8: 38415905-38415906 on Build GRCh38
rs377314381 Chr.8: 38415915-38415915 on Build GRCh38
rs751840290 Chr.8: 38415949-38415949 on Build GRCh38
rs781268209 Chr.8: 38415952-38415952 on Build GRCh38
rs374740971 Chr.8: 38415909-38415909 on Build GRCh38
rs947526102 Chr.8: 38415858-38415858 on Build GRCh38
rs121909629 Chr.8: 38415905-38415905 on Build GRCh38
rs745533062 Chr.8: 38415853-38415853 on Build GRCh38
rs121909639 Chr.8: 38415899-38415899 on Build GRCh38
rs749160589 Chr.8: 38415960-38415960 on Build GRCh38
rs779555140 Chr.8: 38415906-38415907 on Build GRCh38
rs777647992 Chr.8: 38415844-38415844 on Build GRCh38
rs374046152 Chr.8: 38415847-38415847 on Build GRCh38

View Full Product Details