Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41260590-41260863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UCHL1
Entrez Gene ID
7345
Gene Name
ubiquitin C-terminal hydrolase L1
Gene Aliases
HEL-117, HEL-S-53, NDGOA, PARK5, PGP 9.5, PGP9.5, PGP95, Uch-L1
UniGene
Hs.518731
Gene Chromosome Location
Chr.4: 41256881-41268429 on Build GRCh38
Transcripts
NM_004181.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5964817 c.325+3A>G p.? Chr.4: 41260800-41260800 on Build GRCh38
4332740 c.299C>A p.A100D Chr.4: 41260771-41260771 on Build GRCh38
40738 c.298G>A p.A100T Chr.4: 41260770-41260770 on Build GRCh38
5866397 c.289C>T p.H97Y Chr.4: 41260761-41260761 on Build GRCh38
4839209 c.181A>C p.N61H Chr.4: 41260653-41260653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs79021365 Chr.4: 41260629-41260629 on Build GRCh38
rs770737600 Chr.4: 41260639-41260639 on Build GRCh38
rs373327947 Chr.4: 41260642-41260642 on Build GRCh38
rs774414437 Chr.4: 41260821-41260822 on Build GRCh38
rs11556275 Chr.4: 41260708-41260708 on Build GRCh38
rs773363300 Chr.4: 41260619-41260619 on Build GRCh38
rs189371541 Chr.4: 41260687-41260687 on Build GRCh38
rs369024443 Chr.4: 41260804-41260804 on Build GRCh38
rs778240876 Chr.4: 41260643-41260643 on Build GRCh38
rs762747139 Chr.4: 41260830-41260830 on Build GRCh38
rs769407848 Chr.4: 41260816-41260816 on Build GRCh38
rs148096293 Chr.4: 41260695-41260695 on Build GRCh38
rs751276790 Chr.4: 41260664-41260664 on Build GRCh38
rs747330895 Chr.4: 41260698-41260698 on Build GRCh38
rs910408396 Chr.4: 41260672-41260672 on Build GRCh38
rs748331745 Chr.4: 41260631-41260631 on Build GRCh38
rs369933510 Chr.4: 41260630-41260630 on Build GRCh38
rs745686726 Chr.4: 41260808-41260808 on Build GRCh38
rs756641317 Chr.4: 41260709-41260709 on Build GRCh38
rs759432898 Chr.4: 41260640-41260640 on Build GRCh38
rs121917767 Chr.4: 41260751-41260751 on Build GRCh38
rs775527627 Chr.4: 41260818-41260818 on Build GRCh38
rs192697162 Chr.4: 41260763-41260763 on Build GRCh38
rs754234212 Chr.4: 41260746-41260746 on Build GRCh38
rs780627453 Chr.4: 41260715-41260715 on Build GRCh38
rs772252224 Chr.4: 41260786-41260786 on Build GRCh38
rs761532352 Chr.4: 41260671-41260671 on Build GRCh38
rs11556279 Chr.4: 41260738-41260738 on Build GRCh38
rs748417996 Chr.4: 41260764-41260764 on Build GRCh38
rs111649115 Chr.4: 41260736-41260736 on Build GRCh38
rs908612470 Chr.4: 41260829-41260829 on Build GRCh38

View Full Product Details