Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27920328-27920553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDX1
Entrez Gene ID
3651
Gene Name
pancreatic and duodenal homeobox 1
Gene Aliases
GSF, IDX-1, IPF1, IUF1, MODY4, PAGEN1, PDX-1, STF-1
UniGene
Hs.32938
Gene Chromosome Location
Chr.13: 27919982-27926314 on Build GRCh38
Transcripts
NM_000209.3 NR_047484.1

Gene Symbol
PDX1-AS1
Entrez Gene ID
100861550
Gene Name
PDX1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.416201
Gene Chromosome Location
Chr.13: 27829759-27921404 on Build GRCh38
Transcripts
NM_000209.3 NR_047484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4543055 c.330G>A p.E110E Chr.13: 27920468-27920468 on Build GRCh38
1706591 c.353C>T p.P118L Chr.13: 27920491-27920491 on Build GRCh38
2070076 c.385G>A p.A129T Chr.13: 27920523-27920523 on Build GRCh38
6400213 c.311C>T p.A104V Chr.13: 27920449-27920449 on Build GRCh38
3793231 c.331G>A p.E111K Chr.13: 27920469-27920469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760443155 Chr.13: 27920479-27920479 on Build GRCh38
rs765919889 Chr.13: 27920418-27920418 on Build GRCh38
rs756909200 Chr.13: 27920414-27920416 on Build GRCh38
rs987911104 Chr.13: 27920381-27920381 on Build GRCh38
rs927606533 Chr.13: 27920419-27920419 on Build GRCh38
rs890309056 Chr.13: 27920498-27920498 on Build GRCh38
rs752236361 Chr.13: 27920369-27920369 on Build GRCh38
rs772939313 Chr.13: 27920411-27920411 on Build GRCh38
rs957995159 Chr.13: 27920377-27920377 on Build GRCh38
rs572689574 Chr.13: 27920353-27920353 on Build GRCh38
rs778651779 Chr.13: 27920417-27920418 on Build GRCh38
rs757558416 Chr.13: 27920533-27920533 on Build GRCh38
rs752260094 Chr.13: 27920438-27920438 on Build GRCh38
rs753499583 Chr.13: 27920410-27920411 on Build GRCh38
rs745911015 Chr.13: 27920396-27920396 on Build GRCh38
rs769974289 Chr.13: 27920397-27920397 on Build GRCh38
rs772008981 Chr.13: 27920409-27920409 on Build GRCh38
rs149275902 Chr.13: 27920474-27920474 on Build GRCh38
rs749389364 Chr.13: 27920471-27920471 on Build GRCh38
rs780653054 Chr.13: 27920438-27920438 on Build GRCh38
rs375009107 Chr.13: 27920422-27920422 on Build GRCh38
rs797045885 Chr.13: 27920423-27920423 on Build GRCh38
rs762584321 Chr.13: 27920516-27920516 on Build GRCh38
rs193922353 Chr.13: 27920354-27920354 on Build GRCh38
rs781552927 Chr.13: 27920534-27920534 on Build GRCh38
rs752207066 Chr.13: 27920507-27920507 on Build GRCh38
rs193922354 Chr.13: 27920456-27920456 on Build GRCh38
rs758841244 Chr.13: 27920426-27920427 on Build GRCh38
rs201701313 Chr.13: 27920459-27920459 on Build GRCh38
rs764810731 Chr.13: 27920502-27920502 on Build GRCh38
rs764675125 Chr.13: 27920522-27920522 on Build GRCh38
rs1037129874 Chr.13: 27920458-27920458 on Build GRCh38
rs778447909 Chr.13: 27920441-27920441 on Build GRCh38
rs760498179 Chr.13: 27920416-27920416 on Build GRCh38
rs758919455 Chr.13: 27920440-27920440 on Build GRCh38
rs781516081 Chr.13: 27920385-27920385 on Build GRCh38
rs764870332 Chr.13: 27920434-27920434 on Build GRCh38
rs925558250 Chr.13: 27920400-27920400 on Build GRCh38
rs901280387 Chr.13: 27920460-27920460 on Build GRCh38
rs748725637 Chr.13: 27920348-27920348 on Build GRCh38
rs778216763 Chr.13: 27920352-27920352 on Build GRCh38
rs546786946 Chr.13: 27920399-27920399 on Build GRCh38
rs757687785 Chr.13: 27920445-27920445 on Build GRCh38
rs998120986 Chr.13: 27920462-27920462 on Build GRCh38
rs751940952 Chr.13: 27920442-27920442 on Build GRCh38
rs759162149 Chr.13: 27920489-27920489 on Build GRCh38
rs752168064 Chr.13: 27920532-27920532 on Build GRCh38
rs566665917 Chr.13: 27920413-27920413 on Build GRCh38
rs535722487 Chr.13: 27920450-27920450 on Build GRCh38
rs564129447 Chr.13: 27920349-27920349 on Build GRCh38
rs1046482148 Chr.13: 27920426-27920426 on Build GRCh38
rs573752557 Chr.13: 27920487-27920487 on Build GRCh38
rs773176417 Chr.13: 27920476-27920476 on Build GRCh38
rs137852783 Chr.13: 27920364-27920364 on Build GRCh38
rs746180578 Chr.13: 27920422-27920423 on Build GRCh38

View Full Product Details