Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50581519-50581792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHKB
Entrez Gene ID
1120
Gene Name
choline kinase beta
Gene Aliases
CHETK, CHKL, CK, CKB, CKEKB, EK, EKB, MDCMC
UniGene
Hs.654827
Gene Chromosome Location
Chr.22: 50578958-50582999 on Build GRCh38
Transcripts
NM_005198.4 NR_027928.2

Gene Symbol
CHKB-CPT1B
Entrez Gene ID
386593
Gene Name
CHKB-CPT1B readthrough (NMD candidate)
Gene Aliases
CHKL-CPT1B
UniGene
Hs.439777
Gene Chromosome Location
Chr.22: 50568861-50582999 on Build GRCh38
Transcripts
NM_005198.4 NR_027928.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1308298 c.475C>T p.R159* Chr.22: 50581526-50581526 on Build GRCh38
6628414 c.443T>C p.I148T Chr.22: 50581753-50581753 on Build GRCh38
3555812 c.404T>C p.L135P Chr.22: 50581792-50581792 on Build GRCh38
118574 c.451C>A p.R151R Chr.22: 50581550-50581550 on Build GRCh38
1035539 c.464C>A p.T155N Chr.22: 50581537-50581537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777875627 Chr.22: 50581709-50581709 on Build GRCh38
rs1017133958 Chr.22: 50581753-50581753 on Build GRCh38
rs762694630 Chr.22: 50581771-50581771 on Build GRCh38
rs932996910 Chr.22: 50581712-50581712 on Build GRCh38
rs201257544 Chr.22: 50581768-50581768 on Build GRCh38
rs148857303 Chr.22: 50581550-50581550 on Build GRCh38
rs927411775 Chr.22: 50581644-50581644 on Build GRCh38
rs1014513349 Chr.22: 50581689-50581689 on Build GRCh38
rs749454476 Chr.22: 50581737-50581737 on Build GRCh38
rs572783600 Chr.22: 50581605-50581605 on Build GRCh38
rs750048050 Chr.22: 50581554-50581554 on Build GRCh38
rs981813967 Chr.22: 50581641-50581641 on Build GRCh38
rs786205117 Chr.22: 50581542-50581543 on Build GRCh38
rs1050486484 Chr.22: 50581716-50581716 on Build GRCh38
rs765924917 Chr.22: 50581592-50581592 on Build GRCh38
rs914393061 Chr.22: 50581681-50581681 on Build GRCh38
rs888889702 Chr.22: 50581731-50581731 on Build GRCh38
rs370952926 Chr.22: 50581700-50581700 on Build GRCh38
rs146693439 Chr.22: 50581544-50581544 on Build GRCh38
rs961952098 Chr.22: 50581692-50581692 on Build GRCh38
rs371067691 Chr.22: 50581744-50581744 on Build GRCh38
rs918626326 Chr.22: 50581607-50581607 on Build GRCh38
rs991694422 Chr.22: 50581747-50581747 on Build GRCh38
rs62239539 Chr.22: 50581742-50581742 on Build GRCh38
rs777435553 Chr.22: 50581584-50581585 on Build GRCh38
rs946000794 Chr.22: 50581682-50581682 on Build GRCh38
rs373931091 Chr.22: 50581581-50581581 on Build GRCh38
rs890161153 Chr.22: 50581674-50581674 on Build GRCh38
rs901728719 Chr.22: 50581711-50581711 on Build GRCh38
rs900029985 Chr.22: 50581631-50581631 on Build GRCh38
rs771001653 Chr.22: 50581739-50581739 on Build GRCh38
rs1054695441 Chr.22: 50581606-50581606 on Build GRCh38
rs2269381 Chr.22: 50581640-50581640 on Build GRCh38
rs113158900 Chr.22: 50581702-50581702 on Build GRCh38
rs997415280 Chr.22: 50581632-50581632 on Build GRCh38
rs184839240 Chr.22: 50581549-50581549 on Build GRCh38
rs368201426 Chr.22: 50581703-50581703 on Build GRCh38
rs776594113 Chr.22: 50581756-50581756 on Build GRCh38
rs757967618 Chr.22: 50581585-50581605 on Build GRCh38
rs375161469 Chr.22: 50581745-50581745 on Build GRCh38
rs773932223 Chr.22: 50581680-50581680 on Build GRCh38
rs549595714 Chr.22: 50581704-50581704 on Build GRCh38
rs761535800 Chr.22: 50581758-50581758 on Build GRCh38
rs917349763 Chr.22: 50581752-50581752 on Build GRCh38
rs937396896 Chr.22: 50581665-50581665 on Build GRCh38
rs374563842 Chr.22: 50581600-50581600 on Build GRCh38

View Full Product Details