Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 49146713-49146986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOLH1
Entrez Gene ID
2346
Gene Name
folate hydrolase (prostate-specific membrane antigen) 1
Gene Aliases
FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP
UniGene
Hs.645352
Gene Chromosome Location
Chr.11: 49145094-49208706 on Build GRCh38
Transcripts
NM_001014986.1 NM_001193471.1 NM_001193472.1 NM_001193473.1 NM_004476.1 XM_017017432.1 XM_017017433.1 XM_017017434.1 XM_017017435.1 XM_017017436.1 XM_017017437.1 XM_017017438.1 XM_017017439.1 XM_017017440.1 XM_017017441.1 XM_017017442.1 XM_017017443.1 XM_017017444.1 XM_017017445.1 XM_017017446.1 XM_017017447.1 XM_017017448.1 XM_017017449.1 XM_017017450.1 XM_017017451.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3869274 c.2120G>A p.G707E Chr.11: 49146889-49146889 on Build GRCh38
4768768 c.2154A>C p.K718N Chr.11: 49146855-49146855 on Build GRCh38
3782631 c.2097G>C p.K699N Chr.11: 49146912-49146912 on Build GRCh38
5773415 c.2238G>C p.L746F Chr.11: 49146771-49146771 on Build GRCh38
4974994 c.2065C>T p.H689Y Chr.11: 49146944-49146944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753409936 Chr.11: 49146857-49146857 on Build GRCh38
rs868659595 Chr.11: 49146889-49146889 on Build GRCh38
rs149464210 Chr.11: 49146960-49146960 on Build GRCh38
rs149242168 Chr.11: 49146922-49146922 on Build GRCh38
rs756160678 Chr.11: 49146833-49146833 on Build GRCh38
rs763102212 Chr.11: 49146915-49146915 on Build GRCh38
rs150088430 Chr.11: 49146858-49146858 on Build GRCh38
rs368721018 Chr.11: 49146908-49146908 on Build GRCh38
rs963654912 Chr.11: 49146936-49146936 on Build GRCh38
rs1803128 Chr.11: 49146828-49146828 on Build GRCh38
rs752660381 Chr.11: 49146810-49146810 on Build GRCh38
rs748619991 Chr.11: 49146875-49146875 on Build GRCh38
rs773315602 Chr.11: 49146751-49146751 on Build GRCh38
rs560179737 Chr.11: 49146859-49146859 on Build GRCh38
rs1803127 Chr.11: 49146811-49146811 on Build GRCh38
rs550087931 Chr.11: 49146766-49146766 on Build GRCh38
rs770985396 Chr.11: 49146918-49146918 on Build GRCh38
rs201855858 Chr.11: 49146864-49146864 on Build GRCh38
rs766640846 Chr.11: 49146759-49146759 on Build GRCh38
rs767390691 Chr.11: 49146959-49146959 on Build GRCh38
rs770312515 Chr.11: 49146902-49146902 on Build GRCh38
rs763040665 Chr.11: 49146754-49146754 on Build GRCh38
rs759943723 Chr.11: 49146793-49146793 on Build GRCh38
rs595139 Chr.11: 49146748-49146748 on Build GRCh38
rs781582032 Chr.11: 49146866-49146866 on Build GRCh38
rs759372952 Chr.11: 49146796-49146796 on Build GRCh38
rs777377989 Chr.11: 49146840-49146840 on Build GRCh38
rs140860006 Chr.11: 49146799-49146799 on Build GRCh38
rs112972107 Chr.11: 49146919-49146919 on Build GRCh38
rs767480479 Chr.11: 49146802-49146802 on Build GRCh38
rs200462865 Chr.11: 49146929-49146929 on Build GRCh38
rs751249618 Chr.11: 49146781-49146781 on Build GRCh38

View Full Product Details