Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9759235-9759508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR143
Entrez Gene ID
4935
Gene Name
G protein-coupled receptor 143
Gene Aliases
NYS6, OA1
UniGene
Hs.74124
Gene Chromosome Location
Chr.X: 9725413-9765965 on Build GRCh38
Transcripts
NM_000273.2 XM_005274541.3

SNP Detail

SNP ID Chromosome Location
rs373296925 Chr.X: 9759434-9759434 on Build GRCh38
rs779583422 Chr.X: 9759319-9759319 on Build GRCh38
rs746567246 Chr.X: 9759435-9759435 on Build GRCh38
rs761647216 Chr.X: 9759340-9759340 on Build GRCh38
rs771486573 Chr.X: 9759323-9759323 on Build GRCh38
rs62635034 Chr.X: 9759385-9759385 on Build GRCh38
rs767508273 Chr.X: 9759350-9759350 on Build GRCh38
rs281865180 Chr.X: 9759342-9759343 on Build GRCh38
rs186055724 Chr.X: 9759343-9759343 on Build GRCh38
rs62635762 Chr.X: 9759374-9759374 on Build GRCh38
rs137852296 Chr.X: 9759390-9759390 on Build GRCh38
rs754062843 Chr.X: 9759376-9759376 on Build GRCh38
rs776232889 Chr.X: 9759329-9759329 on Build GRCh38
rs58933950 Chr.X: 9759332-9759332 on Build GRCh38
rs756145241 Chr.X: 9759357-9759357 on Build GRCh38
rs62635762 Chr.X: 9759374-9759374 on Build GRCh38
rs768418077 Chr.X: 9759327-9759327 on Build GRCh38
rs772688504 Chr.X: 9759426-9759426 on Build GRCh38
rs926231983 Chr.X: 9759489-9759489 on Build GRCh38
rs141573793 Chr.X: 9759424-9759424 on Build GRCh38
rs778099606 Chr.X: 9759372-9759372 on Build GRCh38
rs762815553 Chr.X: 9759344-9759344 on Build GRCh38
rs769585373 Chr.X: 9759477-9759477 on Build GRCh38
rs749739235 Chr.X: 9759300-9759300 on Build GRCh38
rs35528779 Chr.X: 9759387-9759388 on Build GRCh38
rs768027711 Chr.X: 9759439-9759439 on Build GRCh38
rs58068096 Chr.X: 9759347-9759347 on Build GRCh38
rs748188612 Chr.X: 9759325-9759325 on Build GRCh38
rs776290292 Chr.X: 9759455-9759455 on Build GRCh38
rs281865181 Chr.X: 9759331-9759331 on Build GRCh38
rs62635032 Chr.X: 9759416-9759416 on Build GRCh38
rs757567458 Chr.X: 9759472-9759472 on Build GRCh38
rs999252940 Chr.X: 9759270-9759270 on Build GRCh38
rs181078528 Chr.X: 9759312-9759312 on Build GRCh38
rs148358078 Chr.X: 9759391-9759391 on Build GRCh38
rs769824928 Chr.X: 9759459-9759459 on Build GRCh38
rs201830411 Chr.X: 9759351-9759351 on Build GRCh38
rs746336091 Chr.X: 9759322-9759322 on Build GRCh38

View Full Product Details