Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30974366-30974599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHRHR
Entrez Gene ID
2692
Gene Name
growth hormone releasing hormone receptor
Gene Aliases
GHRFR, GRFR, IGHD1B
UniGene
Hs.733003
Gene Chromosome Location
Chr.7: 30964021-30979531 on Build GRCh38
Transcripts
NM_000823.3 XM_011515263.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6670856 c.200G>A p.R67H Chr.7: 30974454-30974454 on Build GRCh38
1622783 c.812C>T p.A271V Chr.7: 30974489-30974489 on Build GRCh38
6670855 c.777G>A p.T259T Chr.7: 30974454-30974454 on Build GRCh38
4951746 c.781G>A p.V261M Chr.7: 30974458-30974458 on Build GRCh38
6424211 c.812C>A p.A271E Chr.7: 30974489-30974489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004753042 Chr.7: 30974435-30974435 on Build GRCh38
rs773383022 Chr.7: 30974502-30974502 on Build GRCh38
rs770406273 Chr.7: 30974424-30974424 on Build GRCh38
rs372209255 Chr.7: 30974436-30974436 on Build GRCh38
rs994762268 Chr.7: 30974464-30974464 on Build GRCh38
rs747780866 Chr.7: 30974406-30974406 on Build GRCh38
rs767254698 Chr.7: 30974465-30974465 on Build GRCh38
rs762086691 Chr.7: 30974463-30974463 on Build GRCh38
rs747823161 Chr.7: 30974487-30974487 on Build GRCh38
rs151020002 Chr.7: 30974501-30974501 on Build GRCh38
rs760163983 Chr.7: 30974421-30974421 on Build GRCh38
rs781024420 Chr.7: 30974385-30974385 on Build GRCh38
rs750425635 Chr.7: 30974390-30974390 on Build GRCh38
rs375167526 Chr.7: 30974511-30974511 on Build GRCh38
rs749787094 Chr.7: 30974496-30974496 on Build GRCh38
rs770246828 Chr.7: 30974490-30974490 on Build GRCh38
rs368435900 Chr.7: 30974402-30974402 on Build GRCh38
rs375562610 Chr.7: 30974437-30974437 on Build GRCh38
rs774492929 Chr.7: 30974453-30974453 on Build GRCh38
rs547906129 Chr.7: 30974489-30974489 on Build GRCh38
rs776149873 Chr.7: 30974494-30974494 on Build GRCh38
rs140869993 Chr.7: 30974473-30974473 on Build GRCh38
rs529712059 Chr.7: 30974482-30974482 on Build GRCh38
rs760469490 Chr.7: 30974503-30974503 on Build GRCh38
rs748818417 Chr.7: 30974397-30974397 on Build GRCh38
rs562517014 Chr.7: 30974478-30974478 on Build GRCh38
rs753708857 Chr.7: 30974479-30974479 on Build GRCh38
rs764172835 Chr.7: 30974440-30974440 on Build GRCh38
rs771259484 Chr.7: 30974408-30974408 on Build GRCh38
rs768172679 Chr.7: 30974401-30974401 on Build GRCh38
rs369516290 Chr.7: 30974519-30974519 on Build GRCh38
rs774456521 Chr.7: 30974500-30974500 on Build GRCh38
rs750194553 Chr.7: 30974468-30974468 on Build GRCh38
rs566313031 Chr.7: 30974497-30974497 on Build GRCh38
rs756012786 Chr.7: 30974392-30974392 on Build GRCh38
rs149678088 Chr.7: 30974488-30974488 on Build GRCh38
rs901243026 Chr.7: 30974546-30974546 on Build GRCh38
rs777134442 Chr.7: 30974416-30974416 on Build GRCh38
rs140873999 Chr.7: 30974554-30974554 on Build GRCh38
rs965923093 Chr.7: 30974550-30974550 on Build GRCh38
rs368859494 Chr.7: 30974454-30974454 on Build GRCh38
rs754940976 Chr.7: 30974520-30974520 on Build GRCh38
rs764696332 Chr.7: 30974531-30974531 on Build GRCh38
rs779588713 Chr.7: 30974396-30974396 on Build GRCh38
rs778474030 Chr.7: 30974484-30974484 on Build GRCh38

View Full Product Details