Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62705254-62705501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSCL2
Entrez Gene ID
26580
Gene Name
BSCL2, seipin lipid droplet biogenesis associated
Gene Aliases
GNG3LG, HMN5, PELD, SPG17
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62709619 on Build GRCh38
Transcripts
NM_001122955.3 NM_001130702.2 NM_032667.6 NR_037948.1 NR_037949.1 NR_037946.1

Gene Symbol
HNRNPUL2-BSCL2
Entrez Gene ID
100534595
Gene Name
HNRNPUL2-BSCL2 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62727384 on Build GRCh38
Transcripts
NM_001122955.3 NM_001130702.2 NM_032667.6 NR_037948.1 NR_037949.1 NR_037946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4968701 c.288G>T p.G96G Chr.11: 62705417-62705417 on Build GRCh38
6995537 c.123G>T p.L41F Chr.11: 62705390-62705390 on Build GRCh38
5507481 c.212+5G>C p.? Chr.11: 62705296-62705296 on Build GRCh38
5507480 c.404+5G>C p.? Chr.11: 62705296-62705296 on Build GRCh38
4199022 c.218C>T p.A73V Chr.11: 62705487-62705487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781626368 Chr.11: 62705429-62705429 on Build GRCh38
rs761180242 Chr.11: 62705320-62705320 on Build GRCh38
rs756668260 Chr.11: 62705435-62705435 on Build GRCh38
rs147314661 Chr.11: 62705406-62705406 on Build GRCh38
rs142016433 Chr.11: 62705372-62705372 on Build GRCh38
rs773037659 Chr.11: 62705329-62705329 on Build GRCh38
rs572867847 Chr.11: 62705432-62705432 on Build GRCh38
rs1057517657 Chr.11: 62705371-62705371 on Build GRCh38
rs148132646 Chr.11: 62705367-62705367 on Build GRCh38
rs747203248 Chr.11: 62705298-62705298 on Build GRCh38
rs557044760 Chr.11: 62705303-62705303 on Build GRCh38
rs200922424 Chr.11: 62705360-62705360 on Build GRCh38
rs781527096 Chr.11: 62705305-62705305 on Build GRCh38
rs759201886 Chr.11: 62705481-62705481 on Build GRCh38
rs776497908 Chr.11: 62705478-62705478 on Build GRCh38
rs957295826 Chr.11: 62705384-62705384 on Build GRCh38
rs953445189 Chr.11: 62705423-62705423 on Build GRCh38
rs776003071 Chr.11: 62705339-62705339 on Build GRCh38
rs867357580 Chr.11: 62705475-62705475 on Build GRCh38
rs765023622 Chr.11: 62705361-62705361 on Build GRCh38
rs786205068 Chr.11: 62705320-62705321 on Build GRCh38
rs767225271 Chr.11: 62705308-62705311 on Build GRCh38
rs771754989 Chr.11: 62705440-62705440 on Build GRCh38
rs990055355 Chr.11: 62705394-62705394 on Build GRCh38
rs1057517658 Chr.11: 62705357-62705358 on Build GRCh38
rs764139253 Chr.11: 62705385-62705385 on Build GRCh38
rs770133194 Chr.11: 62705309-62705309 on Build GRCh38
rs545775232 Chr.11: 62705369-62705369 on Build GRCh38
rs772832629 Chr.11: 62705448-62705448 on Build GRCh38
rs375818964 Chr.11: 62705290-62705290 on Build GRCh38
rs1057517659 Chr.11: 62705320-62705320 on Build GRCh38
rs992580580 Chr.11: 62705291-62705291 on Build GRCh38
rs370905417 Chr.11: 62705346-62705346 on Build GRCh38
rs779111644 Chr.11: 62705285-62705285 on Build GRCh38
rs760380687 Chr.11: 62705335-62705335 on Build GRCh38
rs773357713 Chr.11: 62705319-62705320 on Build GRCh38
rs746458422 Chr.11: 62705308-62705308 on Build GRCh38
rs200598473 Chr.11: 62705286-62705286 on Build GRCh38
rs770641122 Chr.11: 62705458-62705458 on Build GRCh38
rs746722132 Chr.11: 62705456-62705456 on Build GRCh38
rs751840132 Chr.11: 62705397-62705397 on Build GRCh38
rs771883988 Chr.11: 62705327-62705327 on Build GRCh38
rs199982733 Chr.11: 62705336-62705336 on Build GRCh38
rs879253928 Chr.11: 62705433-62705433 on Build GRCh38
rs773926348 Chr.11: 62705318-62705318 on Build GRCh38
rs780387759 Chr.11: 62705439-62705439 on Build GRCh38
rs568354548 Chr.11: 62705420-62705420 on Build GRCh38

View Full Product Details