Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12649792-12650035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAN2B1
Entrez Gene ID
4125
Gene Name
mannosidase alpha class 2B member 1
Gene Aliases
LAMAN, MANB
UniGene
Hs.356769
Gene Chromosome Location
Chr.19: 12646508-12666777 on Build GRCh38
Transcripts
NM_000528.3 NM_001173498.1 XM_005259913.1 XM_017026818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6420386 c.2245C>T p.R749W Chr.19: 12649932-12649932 on Build GRCh38
991368 c.2221C>T p.R741C Chr.19: 12649956-12649956 on Build GRCh38
991370 c.2211G>T p.E737D Chr.19: 12649966-12649966 on Build GRCh38
5432681 c.2163-6G>T p.? Chr.19: 12650020-12650020 on Build GRCh38
1194057 c.2192G>T p.R731L Chr.19: 12649985-12649985 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373539727 Chr.19: 12649865-12649865 on Build GRCh38
rs3815914 Chr.19: 12649850-12649850 on Build GRCh38
rs142210875 Chr.19: 12649999-12649999 on Build GRCh38
rs747176828 Chr.19: 12649916-12649916 on Build GRCh38
rs750890285 Chr.19: 12649972-12649972 on Build GRCh38
rs569077441 Chr.19: 12649810-12649810 on Build GRCh38
rs755197711 Chr.19: 12649998-12649998 on Build GRCh38
rs199700264 Chr.19: 12649986-12649986 on Build GRCh38
rs533770473 Chr.19: 12649887-12649887 on Build GRCh38
rs377717440 Chr.19: 12649884-12649884 on Build GRCh38
rs770976179 Chr.19: 12650014-12650014 on Build GRCh38
rs373240866 Chr.19: 12649931-12649931 on Build GRCh38
rs781197939 Chr.19: 12649927-12649927 on Build GRCh38
rs773182870 Chr.19: 12649872-12649872 on Build GRCh38
rs759094537 Chr.19: 12649885-12649885 on Build GRCh38
rs749183690 Chr.19: 12650013-12650013 on Build GRCh38
rs80338680 Chr.19: 12649932-12649932 on Build GRCh38
rs61234887 Chr.19: 12649959-12649959 on Build GRCh38
rs765753479 Chr.19: 12649966-12649966 on Build GRCh38
rs864621987 Chr.19: 12649946-12649946 on Build GRCh38
rs988476350 Chr.19: 12649826-12649826 on Build GRCh38
rs1003984277 Chr.19: 12649860-12649860 on Build GRCh38
rs370551216 Chr.19: 12649955-12649955 on Build GRCh38
rs766154921 Chr.19: 12649880-12649880 on Build GRCh38
rs762877000 Chr.19: 12649933-12649933 on Build GRCh38
rs750833439 Chr.19: 12649904-12649904 on Build GRCh38
rs544632771 Chr.19: 12649827-12649827 on Build GRCh38
rs757769028 Chr.19: 12649891-12649891 on Build GRCh38
rs886044335 Chr.19: 12649905-12649906 on Build GRCh38
rs143829874 Chr.19: 12650009-12650009 on Build GRCh38
rs138817592 Chr.19: 12649985-12649985 on Build GRCh38
rs769957397 Chr.19: 12650003-12650003 on Build GRCh38
rs774156075 Chr.19: 12649954-12649954 on Build GRCh38
rs141212446 Chr.19: 12649920-12649920 on Build GRCh38
rs201318291 Chr.19: 12650000-12650000 on Build GRCh38
rs376804252 Chr.19: 12649990-12649990 on Build GRCh38
rs761646956 Chr.19: 12649888-12649889 on Build GRCh38
rs572289342 Chr.19: 12649904-12649905 on Build GRCh38
rs761715309 Chr.19: 12649868-12649868 on Build GRCh38
rs1022927322 Chr.19: 12649912-12649912 on Build GRCh38
rs769767974 Chr.19: 12649869-12649869 on Build GRCh38
rs767192781 Chr.19: 12649956-12649956 on Build GRCh38
rs28639634 Chr.19: 12649910-12649910 on Build GRCh38
rs555203186 Chr.19: 12649896-12649896 on Build GRCh38
rs770786458 Chr.19: 12649953-12649953 on Build GRCh38
rs762822402 Chr.19: 12649877-12649877 on Build GRCh38
rs750179985 Chr.19: 12649940-12649943 on Build GRCh38
rs752234339 Chr.19: 12649886-12649886 on Build GRCh38
rs374501850 Chr.19: 12649881-12649881 on Build GRCh38
rs186573686 Chr.19: 12649832-12649832 on Build GRCh38

View Full Product Details