Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9765523-9765762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR143
Entrez Gene ID
4935
Gene Name
G protein-coupled receptor 143
Gene Aliases
NYS6, OA1
UniGene
Hs.74124
Gene Chromosome Location
Chr.X: 9725413-9765965 on Build GRCh38
Transcripts
NM_000273.2 XM_005274541.3

SNP Detail

SNP ID Chromosome Location
rs751927343 Chr.X: 9765737-9765737 on Build GRCh38
rs62635024 Chr.X: 9765586-9765586 on Build GRCh38
rs62635022 Chr.X: 9765643-9765644 on Build GRCh38
rs949566161 Chr.X: 9765684-9765684 on Build GRCh38
rs986147134 Chr.X: 9765600-9765600 on Build GRCh38
rs985314593 Chr.X: 9765631-9765631 on Build GRCh38
rs769943337 Chr.X: 9765607-9765607 on Build GRCh38
rs773515821 Chr.X: 9765612-9765612 on Build GRCh38
rs772540710 Chr.X: 9765546-9765546 on Build GRCh38
rs762032647 Chr.X: 9765601-9765601 on Build GRCh38
rs766780297 Chr.X: 9765626-9765626 on Build GRCh38
rs763020179 Chr.X: 9765616-9765616 on Build GRCh38
rs672601353 Chr.X: 9765594-9765595 on Build GRCh38
rs62635025 Chr.X: 9765585-9765585 on Build GRCh38
rs1055796261 Chr.X: 9765577-9765577 on Build GRCh38
rs925889440 Chr.X: 9765643-9765643 on Build GRCh38
rs62645741 Chr.X: 9765587-9765603 on Build GRCh38
rs1045922605 Chr.X: 9765692-9765692 on Build GRCh38
rs936878316 Chr.X: 9765627-9765627 on Build GRCh38
rs375563631 Chr.X: 9765557-9765557 on Build GRCh38
rs1057518164 Chr.X: 9765648-9765655 on Build GRCh38
rs62635019 Chr.X: 9765702-9765702 on Build GRCh38
rs953538651 Chr.X: 9765599-9765599 on Build GRCh38
rs914094364 Chr.X: 9765637-9765637 on Build GRCh38
rs62635023 Chr.X: 9765643-9765643 on Build GRCh38
rs63749126 Chr.X: 9765625-9765653 on Build GRCh38
rs62635018 Chr.X: 9765714-9765714 on Build GRCh38
rs773956496 Chr.X: 9765545-9765545 on Build GRCh38
rs1052622546 Chr.X: 9765629-9765629 on Build GRCh38
rs962172837 Chr.X: 9765552-9765552 on Build GRCh38
rs747340998 Chr.X: 9765669-9765669 on Build GRCh38
rs62635026 Chr.X: 9765568-9765568 on Build GRCh38
rs281865176 Chr.X: 9765661-9765667 on Build GRCh38
rs772202855 Chr.X: 9765554-9765554 on Build GRCh38
rs895737952 Chr.X: 9765596-9765596 on Build GRCh38
rs1014144695 Chr.X: 9765733-9765733 on Build GRCh38

View Full Product Details