Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159577.1 | NP_001153049.1 | 5 | 1060 | |
NM_001159578.1 | NP_001153050.1 | 6 | 1205 | ||
NM_001159579.1 | NP_001153051.1 | 4 | 836 | ||
NM_001159580.1 | NP_001153052.1 | 4 | 806 | ||
NM_010727.4 | NP_034857.3 | 4 | 836 | ||
GenBank mRNA | AF034745.1 | - | 5 | 1060 | |
AF034746.1 | - | 4 | 764 | ||
AK044127.1 | - | 4 | 789 | ||
AK138326.1 | - | 4 | 807 | ||
AK161194.1 | - | 6 | 1191 | ||
BC040367.1 | - | 4 | 788 | ||
BC139771.1 | - | 4 | 764 | 40111506 | |
KJ418422.1 | - | 4 | 534 |