Gene Transcripts

Gene Symbol
Myom1
Entrez Gene ID
17929
Gene Name
myomesin 1
Gene Aliases
D430047A17Rik
Chromosome Location
Chr. 17: 71019002 - 71126856 on Build GRCm38
UniGene ID
Mm.4103
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083934.1 NP_001077403.1 32 4209
NM_010867.2 NP_034997.2 33 4503
GenBank mRNA AJ012072.1 - 33 4536
Z22866.1 - 33 4482

View Full Product Details