Gene Transcripts

Gene Symbol
Zc3h7b
Entrez Gene ID
20286
Gene Name
zinc finger CCCH type containing 7B
Gene Aliases
Gm627, Scrg3, Zc2h7b, mKIAA1031
Chromosome Location
Chr. 15: 81744848 - 81796269 on Build GRCm38
UniGene ID
Mm.432075
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081016.1 NP_001074485.1 21 2986
GenBank mRNA AK147484.1 - 21 2752
BC059923.1 - 21 2741

View Full Product Details