Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001005525.2 | NP_001005525.1 | 4 | 386 | |
GenBank mRNA | AK005213.1 | - | 2 | 117 | |
AK012697.1 | - | 5 | 453 | ||
AK054360.1 | - | 1 | 956 | ||
AK084745.1 | - | 5 | 433 | ||
AK166941.1 | - | 7 | 624 | ||
BC008229.1 | - | 4 | 344 | ||
BC132550.1 | - | 2 | 170 | 40130827 | |
BC132552.1 | - | 2 | 170 | 40130829 | |
BC145430.1 | - | 2 | 169 | 9053988 | |
BC145431.1 | - | 2 | 170 | ||
BC145432.1 | - | 2 | 170 | ||
CK793427.1 | - | 332 |