Gene Transcripts

Gene Symbol
Tbc1d31
Entrez Gene ID
210544
Gene Name
TBC1 domain family, member 31
Gene Aliases
4B3, D330013L20Rik, Gm85, Wdr67
Chromosome Location
Chr. 15: 57911684 - 57970068 on Build GRCm38
UniGene ID
Mm.390835
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081396.2 NP_001074865.2 17, 18 2610
GenBank mRNA BC050039.1 - 17, 18 2556
BC066830.1 - 17, 18 2542 30463017

View Full Product Details