Gene Transcripts

Gene Symbol
Scfd2
Entrez Gene ID
212986
Gene Name
Sec1 family domain containing 2
Gene Aliases
E430013M20Rik, STXBP1L1
Chromosome Location
Chr. 5: 74204816 - 74531742 on Build GRCm38
UniGene ID
Mm.134147
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114660.2 NP_001108132.1 4 1335
NM_001286437.1 NP_001273366.1 4 1335
NM_178672.7 NP_848787.2 4 1335
GenBank mRNA AK034000.1 - 4 1330
AK036637.1 - 4 1277
AK044408.1 - 4 1314
AK047026.1 - 4 971
AK047738.1 - 4 1278
AK050673.1 - 4 1286
AK054548.1 - 4 1294
AK088353.1 - 4 1221
AK089011.1 - 4 1036
AK135666.1 - 4 571
AK140246.1 - 4 1336
AK143615.1 - 4 1315
AK163028.1 - 4 1294
BC042528.1 - 4 1346 3256631

View Full Product Details