Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164096.1 | NP_001157568.1 | 11, 12 | 984 | |
NM_025298.3 | NP_079574.2 | 12, 13 | 1062 | ||
GenBank mRNA | AK005034.1 | - | 11, 12 | 985 | |
AK012184.1 | - | 12, 13 | 1022 | ||
AK144721.1 | - | 12, 13 | 1026 | ||
AK146279.1 | - | 12, 13 | 1019 | ||
AK159723.1 | - | 12, 13 | 1019 | ||
BC028924.1 | - | 12, 13 | 983 | ||
BC037637.1 | - | 12, 13 | 1012 | 5010681 |