Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178798.3 | NP_848913.1 | 2 | 504 | |
GenBank mRNA | AK030762.1 | - | 3 | 306 | |
AK037437.1 | - | 1 | 1593 | ||
AK046548.1 | - | 2 | 504 | ||
AK049478.1 | - | 2 | 504 | ||
AK053899.1 | - | 2 | 513 | ||
AK088592.1 | - | 2 | 470 | ||
AK157379.1 | - | 2 | 505 | ||
AK164666.1 | - | 2 | 505 | ||
AK165979.1 | - | 2 | 523 | ||
AK202042.1 | - | 1 | 223 | ||
BC038404.1 | - | 1 | 438 | 4949962 |