Gene Transcripts

Gene Symbol
Lca5l
Entrez Gene ID
385668
Gene Name
Leber congenital amaurosis 5-like
Gene Aliases
4921526F01Rik, Lcca5l
Chromosome Location
Chr. 16: 96158405 - 96193993 on Build GRCm38
UniGene ID
Mm.24626
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001492.2 NP_001001492.2 5 814
GenBank mRNA AK029554.1 - 5 808
AK133316.1 - 5 815
BC141285.1 - 4 664 9055897
BC145611.1 - 4 664 9054177

View Full Product Details