Gene Transcripts
- Gene Symbol
- Vmn1r93
- Entrez Gene ID
- 404290
- Gene Name
- vomeronasal 1 receptor 93
- Gene Aliases
- V1rd21
- Chromosome Location
- Chr. 7: 20138123 - 20139088 on Build GRCm38
- UniGene ID
- Mm.377932
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207547.2 |
NP_997430.2
|
1 |
364 |
|
- Gene Symbol
- Gm10665
- Entrez Gene ID
- 100043058
- Gene Name
- predicted gene 10665
- Gene Aliases
- ENSMUSG00000074308, Vmn1r150
- Chromosome Location
- Chr. 7: 22463110 - 22464075 on Build GRCm38
- UniGene ID
- Mm.484986
- Species
- Mus musculus
- Gene Symbol
- Gm10666
- Entrez Gene ID
- 100043037
- Gene Name
- predicted gene 10666
- Gene Aliases
- ENSMUSG00000074310, Vmn1r141
- Chromosome Location
- Chr. 7: 22133776 - 22134741 on Build GRCm38
- UniGene ID
- Mm.484998
- Species
- Mus musculus
- Gene Symbol
- Gm10670
- Entrez Gene ID
- 100042971
- Gene Name
- predicted gene 10670
- Gene Aliases
- ENSMUSG00000074330, Vmn1r102
- Chromosome Location
- Chr. 7: 20473960 - 20474925 on Build GRCm38
- UniGene ID
- Mm.389840
- Species
- Mus musculus
- Gene Symbol
- Gm4175
- Entrez Gene ID
- 100043019
- Gene Name
- predicted gene 4175
- Gene Aliases
- Vmn1r133
- Chromosome Location
- Chr. 7: 21760531 - 21761475 on Build GRCm38
- UniGene ID
- Mm.484994
- Species
- Mus musculus
- Gene Symbol
- Gm4207
- Entrez Gene ID
- 100043069
- Gene Name
- vomeronasal 1 receptor Vmn1r-ps81 pseudogene
- Gene Aliases
- Vmn1r-ps81
- Chromosome Location
- Chr. 7: 22806346 - 22808474 on Build GRCm38
- UniGene ID
- Mm.431911
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC107205.1 |
- |
1 |
363 |
|
BC119491.1 |
- |
|
363 |
|
- Gene Symbol
- Gm4214
- Entrez Gene ID
- 100043079
- Gene Name
- predicted gene 4214
- Gene Aliases
- Vmn1r161
- Chromosome Location
- Chr. 7: 22983729 - 22984673 on Build GRCm38
- UniGene ID
- Mm.482051
- Species
- Mus musculus
- Gene Symbol
- Vmn1r114
- Entrez Gene ID
- 100042996
- Gene Name
- vomeronasal 1 receptor 114
- Gene Aliases
- ENSMUSG00000074323, Gm10669
- Chromosome Location
- Chr. 7: 20811221 - 20812186 on Build GRCm38
- UniGene ID
- Mm.480291
- Species
- Mus musculus
- Gene Symbol
- Vmn1r121
- Entrez Gene ID
- 667240
- Gene Name
- vomeronasal 1 receptor 121
- Gene Aliases
- EG667240, Gm8533
- Chromosome Location
- Chr. 7: 21097569 - 21098513 on Build GRCm38
- UniGene ID
- Mm.480259
- Species
- Mus musculus
- Gene Symbol
- Vmn1r126
- Entrez Gene ID
- 100043013
- Gene Name
- vomeronasal 1 receptor 126
- Gene Aliases
- Gm4170
- Chromosome Location
- Chr. 7: 21300502 - 21301467 on Build GRCm38
- UniGene ID
- Mm.480289
- Species
- Mus musculus
View Full Product Details