Gene Transcripts

Gene Symbol
Suclg2
Entrez Gene ID
20917
Gene Name
succinate-Coenzyme A ligase, GDP-forming, beta subunit
Gene Aliases
AF171077, AW556404, D6Wsu120e, SCS-betaG
Chromosome Location
Chr. 6: 95473009 - 95718846 on Build GRCm38
UniGene ID
Mm.371585
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011507.3 NP_035637.2 9 1073
GenBank mRNA AF058956.1 - 8 915
AK045176.1 - 9 1026
AK156263.1 - 9 1029
AK167136.1 - 9 1028
AK167339.1 - 9 1312
AK172561.1 - 9 1028
AK172633.1 - 9 1028
AK184241.1 - 4 372
BC030947.1 - 1 50
BC043312.1 - 7 768
BC054425.1 - 8 826
BC080781.1 - 9 1028 30470296
EU234009.1 - 9 980

View Full Product Details