Gene Transcripts
- Gene Symbol
- Ttyh1
- Entrez Gene ID
- 57776
- Gene Name
- tweety homolog 1 (Drosophila)
- Gene Aliases
- 4930459B04Rik, 6330408P11Rik, TTY1, mTTY1, tty
- Chromosome Location
- Chr. 7: 4119527 - 4136241 on Build GRCm38
- UniGene ID
- Mm.29729
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB162929.1 |
- |
5, 6 |
742 |
|
AF190699.1 |
- |
5, 6 |
814 |
|
AF190991.1 |
- |
5, 6 |
814 |
|
AK015486.1 |
- |
4, 5 |
465 |
|
AK018148.1 |
- |
5, 6 |
806 |
|
AK031550.1 |
- |
5, 6 |
842 |
|
AK042758.1 |
- |
5, 6 |
595 |
|
AK043998.1 |
- |
5, 6 |
862 |
|
AK048413.1 |
- |
5, 6 |
813 |
|
AK049903.1 |
- |
4, 5 |
548 |
|
AK081981.1 |
- |
5, 6 |
816 |
|
AK083126.1 |
- |
4, 5 |
939 |
|
BC046310.1 |
- |
5, 6 |
800 |
6307420 |
BC065694.1 |
- |
5, 6 |
806 |
6813365 |
View Full Product Details