Gene Transcripts

Gene Symbol
Cnppd1
Entrez Gene ID
69171
Gene Name
cyclin Pas1/PHO80 domain containing 1
Gene Aliases
1810031K17Rik, AU043960
Chromosome Location
Chr. 1: 75135215 - 75142733 on Build GRCm38
UniGene ID
Mm.28193
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026977.2 NP_081253.1 2 384
GenBank mRNA AK029090.1 - 2 384
AK145962.1 - 3 631
AK159252.1 - 106
AK169779.1 - 2 656
BC013714.1 - 1 157
BC028815.1 - 2 369 5126467

View Full Product Details